EMAIL THIS PAGE TO A FRIEND
Nature genetics
Recurrent PAX3-MAML3 fusion in biphenotypic sinonasal sarcoma.
PMID 24859338
Abstract
Biphenotypic sinonasal sarcoma (SNS) is a newly described tumor of the nasal and paranasal areas. Here we report a recurrent chromosomal translocation in SNS, t(2;4)(q35;q31.1), resulting in a PAX3-MAML3 fusion protein that is a potent transcriptional activator of PAX3 response elements. The SNS phenotype is characterized by aberrant expression of genes involved in neuroectodermal and myogenic differentiation, closely simulating the developmental roles of PAX3.