EMAIL THIS PAGE TO A FRIEND
International journal of oncology
MST4 promotes hepatocellular carcinoma epithelial-mesenchymal transition and metastasis via activation of the p-ERK pathway.
PMID 24859810
Abstract
Mammalian sterile-20-like kinase 4 (MST4) has been implicated in cell proliferation and differentiation. In a previous study, we found MST4 to be an important candidate gene for metastatic hepatocellular carcinoma (HCC); however, the molecular mechanism of the promoting role of MST4 in HCC metastasis is poorly understood. In this study, we show that high expression of MST4 was detected in highly invasive HCC cells and in human HCC specimens with vascular invasion. A high level of MST4, associated with large tumor size, microvascular invasion, presence of intrahepatic metastasis, and advanced TNM classification of malignant tumors stage, was an independent prognostic factor for overall survival (P=0.004) and time to recurrence (P=0.001) after hepatectomy. Knockdown of MST4 expression in HCC cells inhibited cell proliferation, colony formation, and invasion, whereas upregulation of MST4 significantly promoted these processes by promoting epithelial-mesenchymal transition (EMT), dependent on the activation of extracellular signal-regulated protein kinase (ERK) signaling pathways. Furthermore, the combination of MST4 and phosphorylated ERK was proven to have more power to predict the outcomes of HCC patients. This study presents clinical evidence for predicting the value of MST4 in HCC overall survival and recurrence and describes the key role of MST4 in facilitating the EMT process via regulating the activation of ERK, indicating its potential role as a target for postoperative adjuvant therapy for HCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
C7855
Monoclonal Anti-Cdh1 antibody produced in mouse, ~2 mg/mL, clone DCS-266, purified immunoglobulin, buffered aqueous solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution