EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
ArsH from Synechocystis sp. PCC 6803 reduces chromate and ferric iron.
PMID 24861149
Abstract
ArsH is widely distributed in bacteria, and its function remains to be characterized. In this study, we investigated the function of ArsH from Synechocystis sp. PCC 6803. The inactivation of arsH by insertion of a kanamycin-resistance gene in Synechocystis sp. PCC 6803 resulted in the decrease of arsenic and chromium accumulation compared with the wild type. ArsH expression in Escherichia coli strain Rosetta increased its resistance to chromate by reducing chromate in the medium and cells to chromium (III). In addition, ArsH in Rosetta conferred resistance to arsenic. The purified Synechocystis ArsH was able to reduce chromate and ferric iron at the expense of NADPH. Nonlinear regression values of K0.5 for chromate and ferric iron were 71.9 ± 17.8 μM and 59.3 ± 13.8 μM, respectively. The expression level of arsH was induced by arsenite and arsenate, but not chromate or ferric iron. Our results suggest that Synechocystis ArsH had no substrate specificities and shared some biochemical properties that other enzymes possessed. ArsH may be involved in coordinating oxidative stress response generated by arsenic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
259225
1,5-Diphenylcarbazide, ACS reagent
C13H14N4O
33152
1,5-Diphenylcarbazide, Reag. Ph. Eur., ≥98.0%, for metal titration
C13H14N4O
D7766
1,5-Diphenylcarbazide, reagent grade
C13H14N4O
27066
Chromium, coating quality, 99.95%, grit
Cr
374849
Chromium, chips, 99.995% trace metals basis
Cr
255610
Chromium, chips, thickness ~2 mm, 99.5%
Cr
229563
Chromium, chunks, ≥99.99% trace metals basis
Cr
266264
Chromium, powder, 99.5%, −100 mesh
Cr
266299
Chromium, powder, ≥99% trace metals basis, −325 mesh
Cr
749052
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr