EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Role of wetland organic matters as photosensitizer for degradation of micropollutants and metabolites.
PMID 24862465
Abstract
Overall photodegradation of pharmaceuticals, personal care products (PPCPs) and pharmaceutical metabolites were investigated in order to evaluate their photochemical fate in aquatic environments in various natural organic matter (NOM) enriched solutions. Tested PPCPs exhibited different rates of loss during direct and indirect photolysis. Here, only ultraviolet (UV) light source was used for direct photolysis and UV together with (3)DOM(*)for indirect photolysis. Diclofenac and sulfamethoxazole were susceptible to photodegradation, whereas carbamazepine, caffeine, paraxanthine and tri(2-chloroethyl) phosphate (TCEP) showed low levels of photodegradation rate, reflecting their conservative photoreactivity. During indirect photodegradation, in contrast to the hydrophilic autochthonous NOM, allochthonous NOM with relatively high molecular weight (MW), specific ultraviolet absorbance (SUVA) and hydrophobicity (e.g., Suwannee River humic acid (SRHA)) revealed to significantly inhibit the photolysis of target micropollutants. The presence of Typha wetland NOM enhanced the indirect photolysis of well-known conservative micopollutants (carbamazepine and paraxanthine). And atenolol, carbamazepine, glimepiride, and N-acetyl-sulfamethoxazole were found to be sensitive to the triplet excited state of dissolved organic matter ((3)DOM(*)) with Typha wetland NOM under deoxygenated condition. This suggests that photolysis in constructed wetlands connected to the wastewater treatment plant can enhance the degradation of some anthropogenic micropollutants by the interaction with (3)DOM(*) in wetlands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284785
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
C14H14O3
W392103
2,4-Hexadienoic acid, ≥99%, FCC
C6H8O2
D4007
5,5-Diphenylhydantoin, ≥99%
C15H12N2O2
A7655
Atenolol, ≥98% (TLC), powder
C14H22N2O3
74827
Atenolol, analytical reference material
C14H22N2O3
1044403
Atenolol, United States Pharmacopeia (USP) Reference Standard
C14H22N2O3
A1340000
Atenolol, European Pharmacopoeia (EP) Reference Standard
C14H22N2O3
C4024
Carbamazepine, powder
C15H12N2O
94496
Carbamazepine, analytical standard
C15H12N2O
C8981
Carbamazepine, meets USP testing specifications
C15H12N2O
C0450000
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
C15H12N2O
1093001
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
C15H12N2O
PHR1067
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O
G2295
Glimepiride, ≥98% (HPLC), solid
C24H34N4O5S
Y0000515
Glimepiride, European Pharmacopoeia (EP) Reference Standard
C24H34N4O5S
1292303
Glimepiride, United States Pharmacopeia (USP) Reference Standard
C24H34N4O5S
PHR1004
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
C13H18O2
I0020000
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
C13H18O2
1335508
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
C13H18O2
Y0000881
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
C13H18O2
N8280
Naproxen, meets USP testing specifications
C14H14O3
46482
Naproxen, VETRANAL, analytical standard
C14H14O3
PHR1040
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
C14H14O3
N0250000
Naproxen, European Pharmacopoeia (EP) Reference Standard
C14H14O3
1457301
Naproxen, United States Pharmacopeia (USP) Reference Standard
C14H14O3
PHR1139
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O2
1535008
Phenytoin, United States Pharmacopeia (USP) Reference Standard
C15H12N2O2
P1290000
Phenytoin, European Pharmacopoeia (EP) Reference Standard
C15H12N2O2
Y0001167
Phenytoin for system suitability, European Pharmacopoeia (EP) Reference Standard
C15H12N2O2
S1626
Sorbic acid, ≥99.0%
C6H8O2