EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development and validation of a stability-indicating LC-UV method for the determination of pantethine and its degradation product based on a forced degradation study.
PMID 24863372
Abstract
Pantethine (d-bis-(N-pantothenyl-β-aminoethyl)-disulfide, PAN), the stable disulfide form of pantetheine, has beneficial effects in vascular diseases being able to decrease the hyperlipidaemia, moderate the platelet function and prevent the lipid peroxidation. Furthermore, recent studies suggested that PAN may be an effective therapeutic agent for cerebral malaria and, possibly, for neurodegenerative processes. Interestingly, in the literature, there were no data dealing with the chemical stability and the analytical aspects of PAN. Hence, in the present work the chemical stability of PAN was for the first time established through a forced degradation study followed by liquid chromatography tandem mass spectrometry investigation showing the formation of three degradation products of PAN (PD1, PD2 and POx) arising from hydrolytic, thermal and oxidative stresses. Based on these data a stability-indicating LC-UV method for simultaneous estimation of PAN, and its most relevant degradation product (PD1) was developed and validated; moreover the method allowed also the separation and the quantification of the preservative system, constituted by a paraben mixture. The method showed linearity for PAN (0.4-1.2mgmL(-1)), MHB, PHB (0.4-1.2μgmL(-1)) and PD1 (2.5-100μgmL(-1)); the precision, determined in terms of intra-day and inter-day precision, expressed as RSDs, were in the ranges 0.4-1.2 and 0.7-1.4, respectively. The method demonstrated to be accurate and robust; indeed the average recoveries were 100.2, 99.9, and 100.0% for PAN, MHB and PHB, respectively, and 99.9% for PD1. By applying small variations of the mobile phase composition, counter-ion concentration and pH the separation of analytes was not affected. Finally, the applicability of this method was evaluated analyzing the available commercial forms at release as well as during stability studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8027
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
CuO4S · 5H2O
C3036
Copper(II) sulfate pentahydrate, plant cell culture tested, ≥98%
CuO4S · 5H2O
203165
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
CuO4S · 5H2O
469130
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
CuO4S · 5H2O
209198
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
CuO4S · 5H2O
C7631
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
CuO4S · 5H2O
31293
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
CuO4S · 5H2O
61245
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
CuO4S · 5H2O
12849
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
CuO4S · 5H2O
RES10395-B7
Cupric sulfate pentahydrate, PharmaGrade, EP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
CuO4S · 5H2O
H5501
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
C8H8O3
H6654
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
C8H8O3
H3647
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
C8H8O3
M8911
Methyl 4-hydroxybenzoate, analytical standard
C8H8O3
54752
Methyl 4-hydroxybenzoate, tested according to Ph.Eur.
C8H8O3
W271004
Methyl 4-hydroxybenzoate, ≥99%, FCC
C8H8O3
47889
Methyl Paraben, analytical standard
C8H8O3
M1650000
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1012
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1432005
Methylparaben, United States Pharmacopeia (USP) Reference Standard
C8H8O3
P5835
Propyl 4-hydroxybenzoate, BioXtra
C10H12O3
54789
Propyl 4-hydroxybenzoate, tested according to Ph.Eur.
C10H12O3
P53357
Propyl 4-hydroxybenzoate, ≥99%
C10H12O3
W295101
Propyl 4-hydroxybenzoate, ≥99%
C10H12O3
P3650000
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C10H12O3
PHR1010
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O3
1577008
Propylparaben, United States Pharmacopeia (USP) Reference Standard
C10H12O3
76954
Sodium 1-pentanesulfonate solution, for ion pair chromatography, concentrate, ampule
C5H11NaO3S
P0299
Sodium pentanesulfonate, ≥95% (elemental analysis)
C5H11NaO3S
P8199
Sodium pentanesulfonate, BioXtra
C5H11NaO3S