EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
S100A8 as potential salivary biomarker of oral squamous cell carcinoma using nanoLC-MS/MS.
PMID 24863804
Abstract
Oral squamous cell carcinoma (OSCC) shows low 5-year survival; early treatment greatly reduces mortality and morbidity. Saliva is a non-invasive sample, with good potential to discover biomarkers for early detection. NanoLC-MS/MS served to analyze saliva proteome from control subjects (n=35) and OSCC patients T1 (n=29), T2 (n=36), T3 (n=14) and T4 (n=21) stages. Identified biomarkers were verified by Western blot and ELISA assays. NanoLC-MS/MS analysis of salivary proteins between 10 and 15kDa identified S100A8, hemoglobin delta and gamma-G globin in T3 and T4 stage OSCC as well as S100A7 in T1 and T2 stage OSCC. Western blot and ELISA indicated positive correlation between salivary S100A8 increment and tumor size stage. High level of S100A8 appeared in 3.4, 13.9, 92.9, and 100% of saliva OSCC patients with T1, T2, T3, and T4 stages, respectively. Significant increase of salivary S100A7 was observed in 20.7% and 11.1% of those with T1 and T2, respectively. AUROC curve indicated high sensitivity, specificity and accuracy of S100A8-based ELISA as a detector. NanoLC-MS/MS, Western blot and ELISA manifested salivary S100A8 as a specific and sensitive marker for detection of OSCC patients. Salivary S100A8 protein could be applicable in developing OSCC diagnostics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501620
Anti-S100A7 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400250
Anti-S100A7 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA006997
Anti-S100A7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408629
Anti-S100A7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411217
Anti-S100A7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2502065
Anti-S100A8 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406407
Anti-S100A8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA024372
Anti-S100A8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411218
Anti-S100A8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411219
Anti-S100A8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0006278M2
Monoclonal Anti-S100A7 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution
SAB1404346
Monoclonal Anti-S100A7 antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution
SAB1409617
Monoclonal Anti-S100A7 antibody produced in mouse, clone 1C5-C6, purified immunoglobulin, buffered aqueous solution
SAB1409618
Monoclonal Anti-S100A7 antibody produced in mouse, clone 3A5, purified immunoglobulin, buffered aqueous solution
SAB1409619
Monoclonal Anti-S100A7 antibody produced in mouse, clone 2A10, purified immunoglobulin, buffered aqueous solution
SAB1409620
Monoclonal Anti-S100A7 antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
SAB1409621
Monoclonal Anti-S100A7 antibody produced in mouse, clone 1F8, purified immunoglobulin, buffered aqueous solution
SAB1409622
Monoclonal Anti-S100A7 antibody produced in mouse, clone 3E11, ascites fluid, solution
WH0006279M1
Monoclonal Anti-S100A8 antibody produced in mouse, clone 2H2, purified immunoglobulin, buffered aqueous solution
SAB1404347
Monoclonal Anti-S100A8 antibody produced in mouse, clone 1B3, purified immunoglobulin, buffered aqueous solution