EMAIL THIS PAGE TO A FRIEND
Cell and tissue research
The receptor for advanced glycation end-products (RAGE) plays a key role in the formation of nanotubes (NTs) between peritoneal mesothelial cells and in murine kidneys.
PMID 24870978
Abstract
The receptor for advanced glycation end-products (RAGE), a multiligand receptor of the immunoglobulin superfamily, takes part in various inflammatory processes. The role of this receptor in the context of intercellular communication, like nanotube (NT)-mediated interaction, is largely unknown. Here, we use cell cultures of human and murine peritoneal mesothelial cells as well as murine kidneys from wild-type and RAGE knockout mouse models to assess the role of RAGE in NT formation and function. We show that loss of RAGE function results in reduced NT numbers under physiological conditions and demonstrate the involvement of MAP kinase signaling in NT formation. Additionally, we show for the first time the existence of NTs in murine kidney tissue and confirm the correlation of RAGE expression and NT numbers. Under elevated oxidative stress conditions like renal ischemia or peritoneal dialysis, we demonstrate that RAGE absence does not prevent NT formation. Rather, increased NT numbers and attenuated kidney tissue damage could be observed, indicating that, depending on the predominant conditions, RAGE affects NT formation with implications for cellular communication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41642
Anti-AGER (AB1) antibody produced in rabbit, affinity isolated antibody
AV41643
Anti-AGER (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2104808
Anti-AGER, (N-terminal) antibody produced in rabbit, affinity isolated antibody
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4300200
Anti-phospho-MAPK14 (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4300201
Anti-phospho-MAPK14 (pTyr182) antibody produced in rabbit, affinity isolated antibody
P1491 Anti-phospho-p38 (pThr180/pTyr182) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504095
Anti-phospho-p38 MAPK (pThr179+Tyr181) antibody produced in rabbit, affinity isolated antibody
SAB4504497
Anti-phospho-p38 MAPK (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4504498
Anti-phospho-p38 MAPK (pTyr182) antibody produced in rabbit, affinity isolated antibody
SAB4504094
Anti-phospho-p38 MAPK (pTyr322) antibody produced in rabbit, affinity isolated antibody
R5278 Anti-RAGE (Receptor for Advanced Glycation Endproducts) antibody produced in rabbit, affinity isolated antibody
SAB1401326
Anti-RAGE antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105049
Anti-RAGE, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW21175B Anti-RK antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
85530
D-Sorbitol, crystallized, ≥99.0% (HPLC)
C6H14O6
97336
D-Sorbitol, liquid, tested according to Ph.Eur.
C6H14O6
M0252
Methylglyoxal solution, ~40% in H2O
C3H4O2
67028
Methylglyoxal solution, technical, ~40% in H2O
C3H4O2
EHU123971 MISSION® esiRNA, esiRNA human RAGE (esiRNA1)
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
W296902
Pyruvaldehyde solution, 40 wt. % in H2O
C3H4O2
S8307
SB 203580, solid, ≥98% (HPLC)
C21H16FN3OS