EMAIL THIS PAGE TO A FRIEND
The Analyst
Rapid analysis of trace volatile formaldehyde in aquatic products by derivatization reaction-based surface enhanced Raman spectroscopy.
PMID 24875278
Abstract
Toxic formaldehyde is sometimes used illegally as a food preservative, however, on-site rapid analysis of trace formaldehyde in aquatic products remains a challenge. In this work, a simple on-site rapid quantification method for trace volatile formaldehyde in aquatic products was developed by a derivative reaction-based surface enhanced Raman spectroscopy (SERS) technique coupled with a homemade portable purge-sampling device. Trace formaldehyde separated from complicated aquatic matrices via a purge-sampling procedure was reacted with a derivative reagent to produce a Raman-active analyte for consequent SERS analysis. Au/SiO2 nanoparticles (NPs) were employed as the enhancement substrate to achieve significant enhancement of Raman signal intensity. Conditions of derivative reaction and SERS detection were optimized in detail, and the selectivity of this analytical method was also evaluated based on related analogs. Under optimal conditions, an extremely low detection limit of 0.17 μg L(-1) was achieved. Trace volatile formaldehyde can be found in fresh squid and shrimp samples without obvious matrix interference, and this was quantified to be 0.13-0.21 mg kg(-1) using the described method. The recoveries of spiked aquatic product samples were found to be 70.0-89.1% with RSDs of 2.3-7.2% (n = 3). The results suggest that the proposed method is reliable and suitable for on-site rapid analysis of trace formaldehyde in aquatic products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00070
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
C2H4O
402788
Acetaldehyde, ACS reagent, ≥99.5%
C2H4O
00071
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
C2H4O
W200336
Acetaldehyde, natural, ≥99%, FG
C2H4O
W200301
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
W200360
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
110078
Acetaldehyde, ReagentPlus®, 99%
C2H4O
W200320
Acetaldehyde, ≥99%, FCC, stabilized
C2H4O
506788
Acetaldehyde, analytical standard
C2H4O
W200379
Acetaldehyde solution, 40 wt. % in H2O
C2H4O
W200387
Acetaldehyde solution, 40 wt. % in isopropanol
C2H4O
W200344
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
C2H4O
W200328
Acetaldehyde solution, 50 wt. % in ethanol
C2H4O
719099
Acetaldehyde solution, 5 M in THF
C2H4O
W200300
Acetaldehyde solution, natural, 50 wt. % in ethanol
C2H4O
94194
Butyraldehyde, analytical standard, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabilizer
C4H8O
418102
Butyraldehyde, purified by redistillation, ≥99.5%
C4H8O
538191
Butyraldehyde, ≥99.0%, dry
C4H8O
W221902
Butyraldehyde, ≥96.0%
C4H8O
W221910
Butyraldehyde, natural, FG
C4H8O
115606
Hexanal, 98%
C6H12O
W255718
Hexanal, ≥97%, FCC, FG
C6H12O
W255726
Hexanal, natural, ≥95%, FG
C6H12O
18109
Hexanal, analytical standard
C6H12O
W292303
Propionaldehyde, ≥97%, FG
C3H6O
538124
Propionaldehyde, reagent grade, 97%
C3H6O
64409
Propionaldehyde, analytical standard
C3H6O