EMAIL THIS PAGE TO A FRIEND
Journal of drug targeting
Polymeric micelles for pH-responsive delivery of cisplatin.
PMID 24878378
Abstract
Methoxy poly(ethylene oxide)-block-poly-(α-carboxylate-ε-caprolactone) (PEO-b-PCCL) was used to develop pH-responsive polymeric micelles for the delivery of cisplatin (CDDP). Micelles were prepared through complexation of CDDP with the pendant carboxyl groups on the poly(ε-caprolactone) core, perhaps through coordinate bonding. The obtained micelles were characterized using dynamic light scattering (DLS) measurement for size and stability. The in vitro release of CDDP at different pHs (7.4, 6.0 and 5.0) was evaluated. The in vitro cell uptake as well as cytotoxicity of developed micelles against two breast cancer cell lines, i.e. MDA-MB-435 and MDA-MB-231, were also assessed and compared to free CDDP as control. DLS results showed PEO-b-PCCL to form stable micelles with an average diameter of <50 nm upon complexation with CDDP. Developed polymeric micelles were capable of slowly releasing CDDP in physiological pH. However, CDDP release from polymeric micelles was triggered upon exposure to electrolytes and/or acidic pHs mimicking that of extracellular tumor microenvironment or intracellular organelles. Consistent with the slow release of CDDP from its polymeric micellar formulation, polymeric micellar CDDP exhibited lower cytotoxicity and CDDP intracellular uptake compared to free drug. The results indicate a great potential for the developed formulation in platinum therapy of breast cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119938
Benzyl chloroformate, ≥98%
C8H7ClO2
386464
Diisopropylamine, purified by redistillation, 99.95%
C6H15N
471224
Diisopropylamine, ≥99.5%
C6H15N
38290
Diisopropylamine, puriss. p.a., ≥99.0% (GC)
C6H15N
38292
Diisopropylamine, analytical standard
C6H15N
704067
ε-Caprolactone, 97%
C6H10O2
52750
Hexane, analytical standard
C6H14
52765
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
C6H14
15671
Hexane, puriss., ≥95% (GC)
C6H14
302120
n-Butyllithium solution, 2.0 M in cyclohexane
C4H9Li
20159
n-Butyllithium solution, 2.7 M in heptane
C4H9Li
230715
n-Butyllithium solution, 11.0 M in hexanes
C4H9Li
230707
n-Butyllithium solution, 2.5 M in hexanes
C4H9Li
186171
n-Butyllithium solution, 1.6 M in hexanes
C4H9Li
302104
n-Butyllithium solution, 2.0 M in pentane
C4H9Li
20160
n-Butyllithium solution, ~1.6 M in hexanes
C4H9Li
710776
n-Butyllithium solution, 1.4 M in toluene
C4H9Li
82648
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
C16H10
18868
Pyrene, certified reference material, TraceCERT®
C16H10
571245
Pyrene, sublimed grade, 99%
C16H10
185515
Pyrene, 98%
C16H10
82650
Pyrene, purum, for fluorescence, ≥97.0% (GC)
C16H10
BCR177R
Pyrene, BCR® certified Reference Material
C16H10
40082
Pyrene, certified reference material, 1000 μg/mL in methanol
C16H10
48570
Pyrene, analytical standard
C16H10