EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Silencing of IFN-stimulated gene transcription is regulated by histone H1 and its chaperone TAF-I.
PMID 24878923
Abstract
Chromatin structure and its alteration play critical roles in the regulation of transcription. However, the transcriptional silencing mechanism with regard to the chromatin structure at an unstimulated state of the interferon (IFN)-stimulated gene (ISG) remains unclear. Here we investigated the role of template activating factor-I (TAF-I, also known as SET) in ISG transcription. Knockdown (KD) of TAF-I increased ISG transcript and simultaneously reduced the histone H1 level on the ISG promoters during the early stages of transcription after IFN stimulation from the unstimulated state. The transcription factor levels on the ISG promoters were increased in TAF-I KD cells only during the early stages of transcription. Furthermore, histone H1 KD also increased ISG transcript. TAF-I and histone H1 double KD did not show the additive effect in ISG transcription, suggesting that TAF-I and histone H1 may act on the same regulatory pathway to control ISG transcription. In addition, TAF-I KD and histone H1 KD affected the chromatin structure near the ISG promoters. On the basis of these findings, we propose that TAF-I and its target histone H1 are key regulators of the chromatin structure at the ISG promoter to maintain the silent state of ISG transcription.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-224
Anti-phospho-STAT2 (Tyr689) Antibody, Detect phospho-STAT2 (Tyr689) also known as signal transducer & activator of transcription 2 with Anti-phospho-STAT2 (Tyr689) Antibody (rabbit polyclonal antibody), that has been demonstrated to work in WB.
SAB4500348
Anti-Histone H3 (Acetyl-Lys14) antibody produced in rabbit, affinity isolated antibody
SAB4500349
Anti-Histone H3 (Acetyl-Lys18) antibody produced in rabbit, affinity isolated antibody
SAB4500350
Anti-Histone H3 (Acetyl-Lys23) antibody produced in rabbit, affinity isolated antibody
SAB4500351
Anti-Histone H3 (Acetyl-Lys27) antibody produced in rabbit, affinity isolated antibody
SAB4500347
Anti-Histone H3 (Acetyl-Lys9) antibody produced in rabbit, affinity isolated antibody
H9289
Anti-Histone H3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
H0164
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406981
Anti-IRF9 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410082
Anti-IRF9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV31200
Anti-ISGF3G antibody produced in rabbit, affinity isolated antibody
HPA001862
ANTI-ISGF3G antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1105169 Anti-STAT1 (671-685) antibody produced in rabbit, IgG fraction of antiserum
SAB4300326
Anti-STAT1 (Ab-701) antibody produced in rabbit, affinity isolated antibody
SAB4300441
Anti-STAT1 (Ab-727) antibody produced in rabbit, affinity isolated antibody
GW22758 Anti-STAT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33373
Anti-STAT1 (AB1) antibody produced in rabbit, affinity isolated antibody
GW22780 Anti-STAT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33374
Anti-STAT1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV38933
Anti-STAT1 (AB3) antibody produced in rabbit, affinity isolated antibody
AV38934
Anti-STAT1 (AB4) antibody produced in rabbit, affinity isolated antibody
SAB1406486
Anti-STAT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000931
Anti-STAT1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36979
Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum
S5683 Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
HPA000982
Anti-STAT1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300591
Anti-STAT2 (Ab-690) antibody produced in rabbit, affinity isolated antibody
HPA018888
Anti-STAT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2