EMAIL THIS PAGE TO A FRIEND
Cancer research
Mast cell-derived prostaglandin D2 inhibits colitis and colitis-associated colon cancer in mice.
PMID 24879565
Abstract
Compared with prostaglandin E2, which has an established role in cancer, the role of the COX metabolite prostaglandin D2 (PGD2) in chronic inflammation leading to tumorigenesis is uncertain. In this study, we investigated the role of PGD2 in colitis and colitis-associated colon cancer (CAC) using genetically modified mice and an established model of inflammatory colon carcinogenesis. Systemic genetic deficiency in hematopoietic PGD synthase (H-PGDS) aggravated colitis and accelerated tumor formation in a manner associated with increased TNFα expression. Treatment with a TNFα receptor antagonist attenuated colitis regardless of genotype. Histologic analysis revealed that infiltrated mast cells strongly expressed H-PGDS in inflamed colons. Mast cell-specific H-PGDS deficiency also aggravated colitis and accelerated CAC. In contrast, treatment with a PGD2 receptor agonist inhibited colitis and CAC. Together, our results identified mast cell-derived PGD2 as an inhibitor of colitis and CAC, with implications for its potential use in preventing or treating colon cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified immunoglobulin, buffered aqueous solution
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution