EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Cardiac glycosides from the bark of Antiaris toxicaria.
PMID 24879902
Abstract
Five new cardiac glycosides (1-5, namely antiaroside Y-ZC) together with 19 known compounds were obtained from the bark of Antiaris toxicaria. Their chemical structures were determined by IR, HR-ESI-MS, 1D and 2D NMR (HSQC, (1)H-(1)H COSY, HMBC, ROESY). The absolute configuration of sugar unit was defined by acid hydrolysis and appropriate derivatization. Compound 1 was rare 5β-H-10β-H-19-nor-cardenolide, which might derive from decarboxylative derivative of 19-COOH cardenolide. The inhibitory effects of cardiac glycosides 1-11 on the viability of NIH-H460 lung cancer cells and their induction of Nur77 expression were evaluated and preliminary structure-activity relationship (SAR) was also discussed.