EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
The effects of the standardized herbal formula PM014 on pulmonary inflammation and airway responsiveness in a murine model of cockroach allergen-induced asthma.
PMID 24879958
Abstract
PM014 is a modified form of the Chung-Sang-Bo-Ha-Tang (CSBHT) herbal formula that has been used to treat chronic pulmonary diseases in Korea for centuries. Previously, we developed a formulation of PM014 based on a series of in vitro and in vivo screening efforts that comprises seven herbal extracts. The PM014 formula includes the root of Rehmannia glutinosa, the cortex of Paeonia suffruticosa, the fruit of Schizandra chinensis, the root of Asparagus cochinchinensis, seeds of Prunus armeniaca, the root of Scutellaria baicalensis and the root of Stemona sessilifolia. Asthma is a chronic inflammatory disease of the lungs that is characterized by wheezing, bronchial contraction, and chest tightness. In addition, the airway becomes hypersensitive and narrows through an inflammatory reaction mediated by Th2 cells. The present study was conducted to evaluate the ability of PM014 to prevent allergic airway inflammation and to attenuate airway responses in a cockroach allergen-induced mouse model. Mice sensitized to and challenged with cockroach allergen were treated with oral administration of PM014. Airway resistance was determined by whole body plethysmography. In addition, Th2 cytokines and immune cell profiles of bronchoalveolar lavage (BAL) fluid and inflammatory mediators in serum were analyzed by ELISA. A series of histological examinations were also conducted to demonstrate the effects of PM014 on airway remodeling, goblet cell hyperplasia and inflammatory responses in the lung. PM014 significantly inhibited the number of total cells, eosinophils, neutrophils, macrophages and lymphocytes in the BAL fluid of mice that were challenged with cockroach allergen. In addition, PM014 reduced the levels of Th2 cytokines (IL-4, IL-5 and IL-13) in the BAL fluid and inflammatory mediators such as IgE in the serum, as measured by enzyme-linked immunosorbent assay (ELISA). Histopathological analysis also showed that PM014 substantially inhibited eosinophil infiltration into the airway, goblet cell hyperplasia and smooth muscle hypertrophy. In this study, our results indicate that PM014 has significant effects on allergic airway inflammation upon exposure to cockroach allergen in a mouse model. According to these outcomes, PM014 may have therapeutic potential as a treatment for allergic asthma.