EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A superfluorinated molecular probe for highly sensitive in vivo(19)F-MRI.
PMID 24884816
Abstract
(19)F-MRI offers unique opportunities to image diseases and track cells and therapeutic agents in vivo. Herein we report a superfluorinated molecular probe, herein called PERFECTA, possessing excellent cellular compatibility, and whose spectral properties, relaxation times, and sensitivity are promising for in vivo (19)F-MRI applications. The molecule, which bears 36 equivalent (19)F atoms and shows a single intense resonance peak, is easily synthesized via a simple one-step reaction and is formulated in water with high stability using trivial reagents and methods.