EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
Comparison of two preclinical myocardial infarct models: coronary coil deployment versus surgical ligation.
PMID 24885652
Abstract
Despite recent advances, myocardial infarction (MI) remains the leading cause of death worldwide. Pre-clinical animal models that closely mimic human MI are pivotal for a quick translation of research and swine have similarities in anatomy and physiology. Here, we compared coronary surgical ligation versus coil embolization MI models in swine. Fifteen animals were randomly distributed to undergo surgical ligation (n=7) or coil embolization (n=8). We evaluated infarct size, scar fibrosis, inflammation, myocardial vascularization, and cardiac function by magnetic resonance imaging (MRI). Thirty-five days after MI, there were no differences between the models in infarct size (P=0.53), left ventricular (LV) ejection fraction (P=0.19), LV end systolic volume (P=0.22), LV end diastolic volume (P=0.84), and cardiac output (P=0.89). Histologically, cardiac scars did not differ and the collagen content, collagen type I (I), collagen type III (III), and the I/III ratio were similar in both groups. Inflammation was assessed using specific anti-CD3 and anti-CD25 antibodies. There was similar activation of inflammation throughout the heart after coil embolization (P=0.78); while, there were more activated lymphocytes in the infarcted myocardium in the surgical occlusion model (P=0.02). Less myocardial vascularization in the infarction areas compared with the border and remote zones only in coil embolization animals was observed (P=0.004 and P=0.014, respectively). Our results support that surgical occlusion and coil embolization MI models generate similar infarct size, cardiac function impairment, and myocardial fibrosis; although, inflammation and myocardial vascularization levels were closer to those found in humans when coil embolization was performed.