PloS one
Soluble CD163 as a marker of macrophage activity in newly diagnosed patients with multiple sclerosis.
PMID 24886843
Abstract
Soluble CD163 (sCD163) is a macrophage specific protein known to be up-regulated in serum from patients with multiple sclerosis (MS). To investigate sCD163 in serum and CSF (cerebrospinal fluid) from patients undergoing MS diagnostic work-up and analyse its potential as a diagnostic biomarker. After a full MS diagnostic work-up, including collection of paired samples of CSF and serum, 183 patients were evaluated for inclusion in this study. Patients were divided into groups based on their diagnosis. Patients with normal clinical and paraclinical findings were grouped as symptomatic controls. Serum and CSF levels of sCD163 were determined by enzyme-linked immunosorbent assay (ELISA). sCD163 could be measured in all serum and CSF samples. A high sCD163 CSF/serum ratio in relation to molecular weight was found, strongly indicating local production in the CNS. Median levels of sCD163 were significantly decreased in serum and significantly elevated in CSF in patients with relapsing-remitting, and primary-progressive MS. There were, however, some overlaps of the measures between groups. In a receiver operating characteristic (ROC) analysis sCD163 CSF/serum ratio had an area under the curve of 0.72. The sCD163 CSF/serum ratio was significantly increased in patients with MS and may reflect macrophage activation in MS lesions. These results suggest that primary progressive MS also is driven by inflammation in which the innate immune system plays a pivotal role.