EMAIL THIS PAGE TO A FRIEND
Bioorganic chemistry
Combined experimental and in silico approaches for exploring antiperoxidative potential of structurally diverse classes of antioxidants on docetaxel-induced lipid peroxidation using 4-HNE as the model marker.
PMID 24887149
Abstract
The objective of the present work was tantamount to explain the antiperoxidative potential and structural requirements of twenty-eight structurally diverse classes of antioxidants on docetaxel-induced lipid peroxidation. Both experimental and computational approaches were taken to the work. The experiments were performed in vitro and goat liver was used as a source of lipid. 4-hydroxy-2-nonenal was used as model marker for estimation of docetaxel-lipid interaction. The computational portion of the work was limited to QSAR analysis of those antioxidants for better understanding of the structural requirements of antioxidants on docetaxel-lipid interaction. The study was done with freely online available 2D descriptors available on PaDEL (open source). Stepwise regression analysis was used as chemometric tool. The experimental study showed the lipid peroxidation induction capacity of docetaxel. It was also noted that all twenty-eight antioxidants had the ability to suppress the lipid peroxidation. But among them butylated hydroxyl toluene showed the highest potential (-20.5%) and flavone showing lowest potential (-0.8%) to suppress the docetaxel-induced lipid peroxidation. The computational study indicates the importance of topology of the whole molecules, topological distances among atoms within a molecule and specific fragment pattern present in a molecule required for inhibition of lipid peroxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52186
(±)-Naringenin, analytical standard
C15H12O5
04732
2,4-Dinitrophenylhydrazine, for HPLC derivatization, moistened with 35% water, ≥99.0% (HPLC)
C6H6N4O4
442361
2-Methylphenol, analytical standard
C7H8O
42251
Apigenin Proteus mirabilis, analytical standard
C15H10O5
01760595
Apigenin Proteus mirabilis, primary pharmaceutical reference standard
C15H10O5
C80105
Chrysin, 97%
C15H10O4
95082
Chrysin, analytical standard
C15H10O4
F4043
Fisetin, ≥98%
C15H10O6 · xH2O
102032
Flavanone, 98%
C15H12O2
F2003
Flavone
C15H10O2
282200
Galangin, autophagy inducing flavonoid
C15H10O5
PHR1136
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8O2
Y0000619
Guaiacol, European Pharmacopoeia (EP) Reference Standard
C7H8O2
H5254
Hesperidin, ≥80%
C28H34O15
50162
Hesperidin, analytical standard
C28H34O15
Y0001203
Hesperidin, European Pharmacopoeia (EP) Reference Standard
C28H34O15
04650590
Hesperidin, primary pharmaceutical reference standard
C28H34O15
96353
Kaempferol, analytical standard
C15H10O6
00550580
Kaempferol, primary pharmaceutical reference standard
C15H10O6
36922
o-Cresol, analytical standard
C7H8O
W348007
o-Cresol, ≥99%
C7H8O
78666
Taxifolin, analytical standard
C15H12O7
03890585
Taxifolin, primary pharmaceutical reference standard
C15H12O7