EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Chemoenzymatic synthesis of functional sialyl Lewis(x) mimetics with a heteroaromatic core.
PMID 24888318
Abstract
Functional mimetics of the sialyl Lewis(X) tetrasaccharide were prepared by the enzymatic sialylation of a 1,3-diglycosylated indole and a glycosyl azide, which was subsequently transformed into a 1,4-diglycosylated 1,2,3-triazole, by using the trans-sialidase of Trypanosoma cruzi. These compounds inhibited the binding of E-, L-, and P-selectin-coated nanoparticles to polyacrylamide-bound sialyl-Lewis(X) -containing neighboring sulfated tyrosine residues (sTyr/sLe(X) -PAA) at low or sub-millimolar concentrations. Except for E-selectin, the mimetics showed higher activities than the natural tetrasaccharide.