EMAIL THIS PAGE TO A FRIEND
Schizophrenia research
A fundamental role for hippocampal parvalbumin in the dopamine hyperfunction associated with schizophrenia.
PMID 24888524
Abstract
Postmortem studies in schizophrenia patients have demonstrated robust alterations in GABAergic markers throughout the neuraxis. It has been suggested that these alterations are restricted to subpopulations of interneurons, such as those containing the calcium binding protein parvalbumin. Indeed, a reduction in parvalbumin expression is a consistent observation in human postmortem studies, as well as, in a wide and diverse variety of animal models. However, it still remains to be determined whether this decrease in parvalbumin expression contributes to, or is a consequence of the disease. Here we utilize lentiviral delivered shRNA and demonstrate that a selective reduction in parvalbumin mRNA expression induces hyperactivity within the ventral hippocampus. In addition, we observe downstream increases in dopamine neuron population activity without changes in average firing rate or percent burst firing. These changes in dopamine neuron activity were associated with an enhanced locomotor response to amphetamine administration. These data therefore demonstrate that a reduction in ventral hippocampal parvalbumin expression is sufficient, in and of itself, to induce an augmented dopamine system function and behavioral hyper-responsivity to amphetamine, implicating a potential key role for parvalbumin in the pathophysiology of schizophrenia.
Related Materials