EMAIL THIS PAGE TO A FRIEND
International journal of laboratory hematology
Automated quantification of apoptosis in B-cell chronic lymphoproliferative disorders: a prognostic variable obtained with the Cell-Dyn Sapphire (Abbott) automated hematology analyzer.
PMID 24888697
Abstract
B-chronic lymphocytic leukemia CLL, a neoplastic clonal disorder with monomorphous small B lymphocytes with scanty cytoplasm and clumped chromatin, can be morphologically differentiated in typical and atypical forms with different prognosis: Smudge cells (Gumprecht's shadows) are one of the well-known features of the typical CLL and are much less inconsistent in other different types CLPD. Abbott Cell-Dyn Sapphire uses the fluorescence after staining with the DNA fluorochrome propidium iodide for the measurement of nucleated red blood cells (NRBCs) and nonviable cells (FL3+ cell fraction): We have studied the possible correlation between presence and number of morphologically identifiable smudge cells on smears and the percentage of nonviable cells produced by Cell-Dyn Sapphire. 305 blood samples from 224 patients with B-cell lymphoproliferative disorders and 40 healthy blood donors were analyzed by CBC performed by Cell-Dyn Sapphire, peripheral blood smear, and immunophenotype characterization. FL3+ fraction in CLPD directly correlated with the percentage of smudge cells and is significantly increased in patients with typical B-CLL. This phenomenon is much less evident in patients with atypical/mixed B-CLL and B-NHL. In small laboratories without FCM and cytogenetic, smudge cells%, can be utilized as a preliminary diagnostic and prognostic tool in differential diagnosis of CLPD.