EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Tanshinone IIA induces cytochrome c-mediated caspase cascade apoptosis in A549 human lung cancer cells via the JNK pathway.
PMID 24888720
Abstract
Tanshinone IIA (TSIIA), a natural diterpene quinone in the traditional Chinese medicinal herb Dan-Shen (Salvia miltiorrhiza), has extensively exerted antitumor activity in cellular and animal models. However, the molecular mechanisms underlying the antitumor effects of TSIIA remain largely unknown. The in vitro effects of TSIIA on apoptosis were investigated in A549 non-small cell lung cancer (NSCLC) cells. The data showed that TSIIA significantly suppressed the proliferation of A549 cells in a dose-dependent manner, with IC50 values of 16.0±3.7 and 14.5±3.3 µM at 48 h as determined by Cell Counting Kit-8 (CCK-8) assay and clone formation assay, respectively. The change of mitochondrial morphology and the loss of mitochondrial membrane potential (MMP) were observed during the induction. Furthermore, TSIIA induced A549 cell apoptosis as confirmed by typical morphological changes, with cytochrome c release from the mitochondria and Bax translocation to the mitochondria. Caspase activity data indicated that TSIIA activated caspase-9 and caspase-3 of mitochondria-mediated apoptosis, but not caspase-8 of receptor-mediated apoptosis, which could be largely rescued by SP600125 (JNK inhibitor). Taken together, these findings provide the first evidence that TSIIA inhibits growth of NSCLC A549 cells, induces activation of JNK signaling and triggers caspase cascade apoptosis mediated by the release of cytochrome c, which provides a better understanding of the molecular mechanisms of TSIIA on lung cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1410987
Anti-CYCS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C5723 Anti-Cytochrome c antibody produced in sheep, whole antiserum
C9616
Anti-Cytochrome c antibody produced in sheep, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
A1293 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2179 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A1682 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7151 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B0529 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5262 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F4018 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F8771 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3682 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701027 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701030 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
A1418 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2429 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A7434
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
B7401 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5387 Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F4143 Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5897 Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9309 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3438 Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution