EMAIL THIS PAGE TO A FRIEND
British journal of cancer
BI-69A11 enhances susceptibility of colon cancer cells to mda-7/IL-24-induced growth inhibition by targeting Akt.
PMID 24892445
Abstract
Akt and its downstream signalling pathways contribute to the aetiology and progression of colorectal carcinoma (CRC). Targeting the Akt pathway is an attractive strategy but few chemotherapeutic drugs have been used to treat CRC with only limited success. BI-69A11, a small molecule inhibitor of Akt, efficiently inhibits growth in melanoma cells. Melanoma differentiation associated gene-7 (mda-7)/interleukin-24 promotes cancer-selective apoptosis when delivered by a tropism-modified replication incompetent adenovirus (Ad.5/3-mda-7). However, Ad.5/3-mda-7 displays diminished antitumour efficacy in several CRC cell lines, which correlates with the expression of K-RAS. The individual and combinatorial effect of BI-69A11 and Ad.5/3-mda-7 in vitro was studied by cell viability, cell cycle, apoptosis and invasion assays in HT29 and HCT116 cells containing wild type or mutant K-ras, respectively. In vivo HT29 tumour xenografts were used to test the efficacy of the combination treatment. BI-69A11 inhibited growth and induced apoptosis in CRC. However, combinatorial treatment was more effective compared with single treatment. This combination showed profound antitumour and anti angiogenic effects in vitro and in vivo by downregulating Akt activity. BI-69A11 enhances the antitumour efficacy of Ad.5/3-mda-7 on CRC overexpressing K-RAS by inducing apoptosis and regulating Akt activity thereby warranting further evaluation in treating CRC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS2301
Anti-AIF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500441
Anti-AIF antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406783
Anti-AIFM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100079
Anti-AIFM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA030611
Anti-AIFM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405469
Anti-BIRC4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411890
ANTI-BIRC4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV40199
Anti-PDCD8 (AB1) antibody produced in rabbit, affinity isolated antibody
AV00028
Anti-PDCD8 (AB2) antibody produced in rabbit, affinity isolated antibody
X4503 Anti-XIAP antibody produced in goat, affinity isolated antibody, lyophilized powder
PRS3331
Anti-XIAP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501453
Anti-XIAP antibody produced in rabbit, affinity isolated antibody
SAB5300151
Monoclonal Anti-AIFM1 antibody produced in mouse, clone 4E7, ascites fluid
WH0000331M1
Monoclonal Anti-BIRC4 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1403557
Monoclonal Anti-BIRC4 antibody produced in mouse, clone 1F4, purified immunoglobulin, buffered aqueous solution