EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Biliary repair and carcinogenesis are mediated by IL-33-dependent cholangiocyte proliferation.
PMID 24892809
Abstract
Injury to the biliary epithelium triggers inflammation and fibrosis, which can result in severe liver diseases and may progress to malignancy. Development of a type 1 immune response has been linked to biliary injury pathogenesis; however, a subset of patients with biliary atresia, the most common childhood cholangiopathy, exhibit increased levels of Th2-promoting cytokines. The relationship among different inflammatory drivers, epithelial repair, and carcinogenesis remains unclear. Here, we determined that the Th2-activating cytokine IL-33 is elevated in biliary atresia patient serum and in the livers and bile ducts of mice with experimental biliary atresia. Administration of IL-33 to WT mice markedly increased cholangiocyte proliferation and promoted sustained cell growth, resulting in dramatic and rapid enlargement of extrahepatic bile ducts. The IL-33-dependent proliferative response was mediated by an increase in the number of type 2 innate lymphoid cells (ILC2s), which released high levels of IL-13 that in turn promoted cholangiocyte hyperplasia. Induction of the IL-33/ILC2/IL-13 circuit in a murine biliary injury model promoted epithelial repair; however, induction of this circuit in mice with constitutive activation of AKT and YAP in bile ducts induced cholangiocarcinoma with liver metastases. These findings reveal that IL-33 mediates epithelial proliferation and suggest that activation of IL-33/ILC2/IL-13 may improve biliary repair and disruption of the circuit may block progression of carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS1105
Anti-CCR3 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS1109
Anti-CCR3 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900012
Anti-CCR3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300654
Anti-CD11c antibody produced in rabbit, affinity isolated antibody
SAB1306027 ANTI-CHK(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405616
Anti-CHKA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV53583
Anti-CHKA antibody produced in rabbit, affinity isolated antibody
HPA024153
Anti-CHKA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410167
Anti-CHKA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1401074
Anti-CHKB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018797
Anti-CHKB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408560
Anti-CHKB antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407519
Anti-CMPK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA053730
Anti-CMPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA058604
Anti-CMPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107657
Anti-CMPK1 antibody produced in rabbit, affinity isolated antibody
SAB1410948
Anti-CMPK1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501670
Anti-Keratin 19 antibody produced in rabbit, affinity isolated antibody
SAB2101302
Anti-KRT19 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101303
Anti-KRT19 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1400148
Anti-KRT19 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA002465 Anti-KRT19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700390 Monoclonal Anti-CD11c antibody produced in mouse, clone BU15, purified immunoglobulin, buffered aqueous solution
SAB4700113
Monoclonal Anti-CD19-PE antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution
SAB4700683 Monoclonal Anti-CD19-PE antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
P7437 Monoclonal Anti-CD19−PE antibody produced in mouse, clone SJ25-C1, purified immunoglobulin, buffered aqueous solution
SAB4700550 Monoclonal Anti-Cd19-PE antibody produced in rat, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB4700474
Monoclonal Anti-CD45-APC antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
SAB4700588
Monoclonal Anti-CD45-APC antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
SAB4700483 Monoclonal Anti-CD45-PE antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution