EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
FTY720 attenuates paraquat-induced lung injury in mice.
PMID 24893116
Abstract
Paraquat (PQ) poisoning, with the lung as a primary target organ, is a devastating disease which irreversibly progresses to diffuse alveolitis followed by extensive lung fibrosis. In the present study, we aimed to investigate the effect of FTY720, an immune modulator, on PQ-induced lung injury in mice. C57BL/6 mice were randomized into four groups: 1) PQ group (n=12): mice was instilled with PQ (30 mg/kg, ip); 2) PQ+FTY720 group (n=12): animals received FTY720 (0.1mg/kg, ip) solution 2h after PQ exposure and twice a week for 4 consecutive weeks; 3) FTY720 group (n=5): FTY720 (0.1mg/kg, ip) was administrated twice a week for 4 consecutive weeks; and 4) Control group (n=10): same volumes of saline were injected. Mice were sacrificed on either day 3 or day 28 for histopathological, biochemical and immunohistochemical analyses of lung damage indicators. We found that FTY720 treatment attenuated PQ-induced acute lung injury and lung fibrosis as evaluated by histopathological changes and Ashcroft score. On day 3, FTY720 administration reduced PQ-induced increases in lung wet weight/body weight (LW/BW), total protein and cytokine levels including interleukin-1β (IL-1β), interleukin-6 (IL-6) and tumor necrosis factor-α (TNF-α) in bronchoalceolar lavage fluid (BALF). On day 28, the expressions of alpha-smooth muscle actin (α-SMA), transforming growth factor-beta (TGF-β) and vascular endothelial growth factor (VEGF) detected by immunohistochemistry, as well as the mRNA levels of α-SMA, Type-I Collagen and Type-III Collagen examined by Real-time PCR were down-regulated after FTY720 treatment. These results indicate that FTY720 could attenuate PQ-induced lung injury, but further investigation is necessary.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2701868
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701827
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701828 Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701689
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701690
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500963
Anti-Smooth Muscle α-Actin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
RAB0306
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0307
Human IL-6 ELISA Kit, for cell and tissue lysates
RAB0313
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
WH0000059M2
Monoclonal Anti-ACTA2 antibody produced in mouse, clone 4A8-2H3, purified immunoglobulin, buffered aqueous solution
SAB1403519
Monoclonal Anti-ACTA2 antibody produced in mouse, clone 1D11-2B9, purified immunoglobulin, buffered aqueous solution
RAB0308
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0309
Mouse IL-6 ELISA Kit, for cell and tissue lysates
RAB0314 Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
RAB0311
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
RAB0312
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate