EMAIL THIS PAGE TO A FRIEND
Nature communications
Homeostatic control of polo-like kinase-1 engenders non-genetic heterogeneity in G2 checkpoint fidelity and timing.
PMID 24893992
Abstract
The G2 checkpoint monitors DNA damage, preventing mitotic entry until the damage can be resolved. The mechanisms controlling checkpoint recovery are unclear. Here, we identify non-genetic heterogeneity in the fidelity and timing of damage-induced G2 checkpoint enforcement in individual cells from the same population. Single-cell fluorescence imaging reveals that individual damaged cells experience varying durations of G2 arrest, and recover with varying levels of remaining checkpoint signal or DNA damage. A gating mechanism dependent on polo-like kinase-1 (PLK1) activity underlies this heterogeneity. PLK1 activity continually accumulates from initial levels in G2-arrested cells, at a rate inversely correlated to checkpoint activation, until it reaches a threshold allowing mitotic entry regardless of remaining checkpoint signal or DNA damage. Thus, homeostatic control of PLK1 by the dynamic opposition between checkpoint signalling and pro-mitotic activities heterogeneously enforces the G2 checkpoint in each individual cell, with implications for cancer pathogenesis and therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, 200 µg, Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, 200 µg, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
05-368
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, 200 µg, Anti-phospho-Ser/Thr-Pro MPM-2 Antibody is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro also known as Mitotic protein #2 & has been tested in ELISA, IHC, IP & WB.
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB2700818
Anti-MDC1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA006915
ANTI-MDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306180 ANTI-PHOSPHO-H3(S10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306179 ANTI-PHOSPHO-H3(S10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305681 ANTI-PHOSPHO-H3(S28) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0000891M1
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1C8, purified immunoglobulin, buffered aqueous solution
SAB5300329 Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1B10, ascites fluid
SAB5300330
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 5G6, ascites fluid
M2444
Monoclonal Anti-MDC1 antibody produced in mouse, clone MDC1-50, purified immunoglobulin, buffered aqueous solution
SAB5200020 Monoclonal Anti-MDC1 antibody produced in mouse, clone P2B11, 1 mg/mL, purified immunoglobulin
PLA0016
Rabbit anti-MDC1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.