EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
TIMP-1 via TWIST1 induces EMT phenotypes in human breast epithelial cells.
PMID 24895412
Abstract
Tissue inhibitor of metalloproteinase-1 (TIMP-1) regulates intracellular signaling networks for inhibition of apoptosis. Tetraspanin (CD63), a cell surface binding partner for TIMP-1, was previously shown to regulate integrin-mediated survival pathways in the human breast epithelial cell line MCF10A. In the current study, we show that TIMP-1 expression induces phenotypic changes in cell morphology, cell adhesion, cytoskeletal remodeling, and motility, indicative of an epithelial-mesenchymal transition (EMT). This is evidenced by loss of the epithelial cell adhesion molecule E-cadherin with an increase in the mesenchymal markers vimentin, N-cadherin, and fibronectin. Signaling through TIMP-1, but not TIMP-2, induces the expression of TWIST1, an important EMT transcription factor known to suppress E-cadherin transcription, in a CD63-dependent manner. RNAi-mediated knockdown of TWIST1 rescued E-cadherin expression in TIMP-1-overexpressing cells, demonstrating a functional significance of TWIST1 in TIMP-1-mediated EMT. Furthermore, analysis of TIMP-1 structural mutants reveals that TIMP-1 interactions with CD63 that activate cell survival signaling and EMT do not require the matrix metalloproteinase (MMP)-inhibitory domain of TIMP-1. Taken together, these data demonstrate that TIMP-1 binding to CD63 activates intracellular signal transduction pathways, resulting in EMT-like changes in breast epithelial cells, independent of its MMP-inhibitory function. TIMP-1's function as an endogenous inhibitor of MMP or as a "cytokine-like" signaling molecule may be a critical determinant for tumor cell behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501702
Anti-TIMP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1409739
Anti-TIMP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4502971
Anti-TIMP1 antibody produced in rabbit, affinity isolated antibody
SAB1411349
Anti-TIMP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406563
Anti-TWIST1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
T6451
Anti-Twist1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV37997
Anti-TWIST1 antibody produced in rabbit, IgG fraction of antiserum
SAB2106420
Anti-TWIST1 antibody produced in rabbit, affinity isolated antibody
SAB1411370
Anti-TWIST1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
EMU037611 MISSION® esiRNA, esiRNA targeting mouse Twist1 (esiRNA1)
EHU151571 MISSION® esiRNA, esiRNA human TWIST1 (esiRNA1)
EHU156431 MISSION® esiRNA, esiRNA human TIMP1 (esiRNA1)
EMU005581 MISSION® esiRNA, esiRNA targeting mouse Timp1 (esiRNA1)
WH0007076M1
Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
SAB1404470 Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4E7, purified immunoglobulin, buffered aqueous solution
WH0007291M1
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
SAB1409772
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4G10, purified immunoglobulin, buffered aqueous solution
SAB1409773 Monoclonal Anti-TWIST1 antibody produced in mouse, clone 1G2, purified immunoglobulin, buffered aqueous solution
SAB1409774
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1409775 Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4G11, purified immunoglobulin, buffered aqueous solution
SAB1409776
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 2G12, purified immunoglobulin, buffered aqueous solution
SAB1409777
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1409778
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3H5, purified immunoglobulin, buffered aqueous solution
SAB1409779
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3A2, purified immunoglobulin, buffered aqueous solution