PloS one
SIRT6 is required for normal retinal function.
PMID 24896097
Abstract
The retina is one of the major energy consuming tissues within the body. In this context, synaptic transmission between light-excited rod and cone photoreceptors and downstream ON-bipolar neurons is a highly demanding energy consuming process. Sirtuin 6 (SIRT6), a NAD-dependent deacylase, plays a key role in regulating glucose metabolism. In this study, we demonstrate that SIRT6 is highly expressed in the retina, controlling levels of histone H3K9 and H3K56 acetylation. Notably, despite apparent normal histology, SIRT6 deficiency caused major retinal transmission defects concomitant to changes in expression of glycolytic genes and glutamate receptors, as well as elevated levels of apoptosis in inner retina cells. Our results identify SIRT6 as a critical modulator of retinal function, likely through its effects on chromatin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV32597
Anti-GRM6 antibody produced in rabbit, affinity isolated antibody
SAB2900137
Anti-GRM6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100976
Anti-GRM6 antibody produced in rabbit, affinity isolated antibody
SAB2104128
Anti-GRM6 antibody produced in rabbit, affinity isolated antibody
S4322
Anti-Sirt6 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500497
Anti-SIRT6 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407499
Anti-SIRT6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV32409
Anti-SIRT6 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2103967
Anti-SIRT6 antibody produced in rabbit, affinity isolated antibody
A1400000
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
C34H46N2O6 · H2O4S · H2O
1045009
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
C34H46N2O6 · H2O4S · H2O
PHR1379
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
C34H46N2O6 · H2O4S · H2O
11332
Atropine sulfate salt monohydrate, tested according to Ph.Eur.
C34H46N2O6 · H2O4S · H2O
A0257
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
C34H46N2O6 · H2O4S · H2O
WH0000060M1
Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution
SAB1403520
Monoclonal Anti-ACTB antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
WH0002916M1
Monoclonal Anti-GRM6 antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution
SAB4200254
Monoclonal Anti-SIRT6 antibody produced in mouse, ~1.0 mg/mL, clone SIRT6-4, purified immunoglobulin
SAB1403256
Monoclonal Anti-SIRT6 antibody produced in mouse, clone 1D8, purified immunoglobulin, buffered aqueous solution