EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and thrombolysis
In vitro thrombolytic activity of purified streptokinase extracted from Streptococcus equinus VIT_VB2 isolated from bovine milk.
PMID 24897956
Abstract
Streptokinase (SK) is an extracellular enzyme secreted by various strains of β-hemolytic Streptococci. The main focus of the current study is to evaluate the in vitro thrombolytic activity of purified SK extracted from Streptococcus equinus VIT_VB2 (Accession no. JX406835) isolated from milk sample. The growth rate of S. equinus VIT_VB2 strain was studied with pH and biomass content which has positive significant effect on enzyme yield. A temperature of 10xa0°C and pH of 6 was found to be optimum for maximum SK activity. The specific activity of the purified SK produced by VIT_VB2 strain was found to be 6,585xa0IUxa0mg(-1). The molecular mass of the enzyme was determined as 47xa0kDa by SDS-PAGE. In vitro thrombolytic activity of purified SK was determined using synthetic chromogenic substrate S-2251, the activity of the purified enzyme was found to be 6,330xa0±xa02.2xa0IU. The purity of SK was compared with standard SK by HPLC. This is the first report which reveals the SK activity of S. equinus isolated from milk sample.