EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Upregulation of the putative oncogene COTE1 contributes to human hepatocarcinogenesis through modulation of WWOX signaling.
PMID 24899407
Abstract
Family with sequence similarity 189, also known as COTE1, has been found to be significantly upregulated in hepatocellular carcinoma (HCC) specimens and cell lines and is associated with tumor size and differentiation. Furthermore, COTE1 contributes to hepatocellular carcinogenesis. The overexpression of COTE1 enhanced in vitro cell viability and colony formation in soft agar, and in vivo tumorigenicity of HCC-derived Focus and Huh7 cells. In contrast, COTE1 knockdown via RNAi markedly suppressed these phenotypes in YY-8103 and WRL-68 HCC cell lines. Mechanistic analyses indicated that COTE1 physically associated with WW domain-containing oxidoreductase (WWOX) and modulated WWOX tyrosine phosphorylation. The ectopic overexpression of COTE1 inhibited the WWOX-p53 signaling pathway by reducing the phosphorylation of WWOX at the Tyr33 residue in Focus cells. Conversely, COTE1 silencing activated tyrosinexa033 phosphorylation of WWOX and induced WWOX-p53 mediated mitochondrial apoptosis in WRL-68 cells. In addition, COTE1 upregulation in Huh7 cells blocked the WWOX-cyclin D1 pathway via dephosphorylation of WWOX Tyr287, stimulating cell cycle progression whereas phosphorylation of Tyr287 of WWOX induced by COTE1 silencing resulted in activation of WWOX-cyclin D1 signaling, leading to cell cycle arrest in YY-8103 cells. Together, our findings suggest that the cytoplasmic protein COTE1 contributes to HCC tumorigenesis by regulating cell proliferation through the modulation of WWOX signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100362
Anti-CCND1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100363
Anti-CCND1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100364
Anti-CCND1 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2106484
Anti-CCND1 antibody produced in rabbit, affinity isolated antibody
C5588 Anti-Cyclin D1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA006659
Anti-FAM189B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2102511
Anti-TP53 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102512
Anti-TP53 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2102513
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2102514
Anti-TP53 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB2102515
Anti-TP53 (ab5) antibody produced in rabbit, affinity isolated antibody
AV02055
Anti-TP53 antibody produced in rabbit, IgG fraction of antiserum
AV37163
Anti-TRP53 antibody produced in rabbit, IgG fraction of antiserum
SAB2106418
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
PLA0072
Rabbit anti-p53 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.