PloS one
BM-1197: a novel and specific Bcl-2/Bcl-xL inhibitor inducing complete and long-lasting tumor regression in vivo.
PMID 24901320
Abstract
Bcl-2 and Bcl-xL are critical regulators of apoptosis that are overexpressed in a variety of human cancers and pharmacological inhibition of Bcl-2 and Bcl-xL represents a promising strategy for cancer treatment. Using a structure-based design approach, we have designed BM-1197 as a potent and efficacious dual inhibitor of Bcl-2 and Bcl-xL. BM-1197 binds to Bcl-2 and Bcl-xL proteins with Ki values less than 1 nM and shows >1,000-fold selectivity over Mcl-1. Mechanistic studies performed in the Mcl-1 knockout mouse embryonic fibroblast (MEF) cells revealed that BM-1197 potently disassociates the heterodimeric interactions between anti-apoptotic and pro-apoptotic Bcl-2 family proteins, concomitant with conformational changes in Bax protein, loss of mitochondrial membrane potential and subsequent cytochrome c release to the cytosol, leading to activation of the caspase cascade and apoptosis. BM-1197 exerts potent growth-inhibitory activity in 7 of 12 small cell lung cancer cell lines tested and induces mechanism-based apoptotic cell death. When intravenously administered at daily or weekly in H146 and H1963 small-cell lung cancer xenograft models, it achieves complete and long-term tumor regression. Consistent with its targeting of Bcl-xL, BM-1197 causes transit platelet reduction in mice. Collectively, our data indicate that BM-1197 is a promising dual Bcl-2/Bcl-xL inhibitor which warrants further investigation as a new anticancer drug.