EMAIL THIS PAGE TO A FRIEND
Nature protocols
In vivo antigen-driven plasmablast enrichment in combination with antigen-specific cell sorting to facilitate the isolation of rare monoclonal antibodies from human B cells.
PMID 24901740
Abstract
The ability to rapidly generate large panels of antigen-specific human antibodies in a rodent would enable the efficient discovery of novel therapeutically useful antibodies. We have developed a system wherein human antigen-specific antibody-secreting plasmablasts can be enriched in vivo, in a severe combined immunodeficient (SCID)/beige mouse host. The antigen-specific plasmablasts can then be sorted by flow cytometry, enabling single-cell cloning and expression of fully human immunoglobulin-G. By using this technique, we have generated four broadly reactive anti-influenza A antibodies. Therefore, the method described here is useful for the identification of rare functional antibodies. This protocol takes ∼1 month to complete, from the time of human vaccination to the cloning of heavy- and light-chain genes. For additional small-scale transient expression, purification and binding analysis, the protocol would take an additional month.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000452
Anti-JUP antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000453
Anti-JUP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41733
Anti-KRT17 antibody produced in rabbit, affinity isolated antibody
HPA043922
Anti-PC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502091
Anti-PC antibody produced in rabbit, affinity isolated antibody
AV44730
Anti-PODXL (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV44729
Anti-PODXL (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2500809
Anti-PODXL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002110
Anti-PODXL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA045507
Anti-PODXL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005550
Anti-PROC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41701
Anti-PROC antibody produced in rabbit, IgG fraction of antiserum
SAB2101884
Anti-PROC antibody produced in rabbit, affinity isolated antibody
SAB2500845
Anti-Pyruvate Carboxylase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9576
Bovine Serum Albumin solution, 30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
WH0003872M1
Monoclonal Anti-KRT17 antibody produced in mouse, clone 2D4-1G9, purified immunoglobulin, buffered aqueous solution
SAB1400147
Monoclonal Anti-KRT17 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
AMAB90644
Monoclonal Anti-PODXL antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0285, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90643
Monoclonal Anti-PODXL antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0284, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90667
Monoclonal Anti-PODXL antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0308, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300033
Monoclonal Anti-PODXL antibody produced in mouse, clone 5D4, ascites fluid
SAB5300034
Monoclonal Anti-PODXL antibody produced in mouse, clone 5F5, ascites fluid
WH0005624M1
Monoclonal Anti-PROC antibody produced in mouse, clone 3A10, purified immunoglobulin, buffered aqueous solution
93283
Tris-EDTA buffer solution, BioUltra, for molecular biology, pH 8.0