EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
PAR1-dependent COX-2/PGE2 production contributes to cell proliferation via EP2 receptors in primary human cardiomyocytes.
PMID 24902855
Abstract
Different protease-activated receptors (PARs) activated by thrombin are involved in cardiovascular disease, via up-regulation of inflammatory proteins including COX-2. However, the mechanisms underlying thrombin-regulated COX-2 expression in human cardiomyocytes remain unclear. Human cardiomyocytes were used in the study. Thrombin-induced COX-2 protein and mRNA expression, and signalling pathways were determined by Western blot, real-time PCR and COX-2 promoter luciferase reporter assays, and pharmacological inhibitors or siRNAs. PGE2 generation and cell proliferation were also determined. Thrombin-induced COX-2 protein and mRNA expression, promoter activity and PGE2 release was attenuated by the PAR1 antagonist (SCH79797) or the inhibitors of proteinase activity (PPACK), MEK1/2 (U0126), p38 MAPK (SB202190) or JNK1/2 (SP600125), and transfection with small interfering RNA (siRNA) of PAR1, p38, p42 or JNK2. These results suggested that PAR1-dependent MAPKs participate in thrombin-induced COX-2 expression in human cardiomyocytes. Moreover, thrombin stimulated phosphorylation of MAPKs, which was attenuated by PPACK and SCH79797. Furthermore, thrombin-induced COX-2 expression was blocked by the inhibitors of AP-1 (tanshinone IIA) and NF-κB (helenalin). Moreover, thrombin-stimulated phosphorylation of c-Jun/AP-1 and p65/NF-κB was attenuated by tanshinone IIA and helenalin, respectively, suggesting that thrombin induces COX-2 expression via PAR1/MAPKs/AP-1 or the NF-κB pathway. Functionally, thrombin increased human cardiomyocyte proliferation through the COX-2/PGE2 system linking to EP2 receptors, as determined by proliferating cell nuclear antigen and cyclin D1 expression. These findings demonstrate that MAPKs-mediated activation of AP-1/NF-κB pathways is, at least in part, required for COX-2/PGE2 /EP2 -triggered cell proliferation in human cardiomyocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1221
AH6809, ≥98%, crystalline solid or supercooled liquid
C17H14O5
SAB2501233
Anti-F2R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405778
Anti-F2R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100740
Anti-F2R antibody produced in rabbit, affinity isolated antibody
SAB2101635
Anti-NR1I2 (ab1) antibody produced in rabbit, affinity isolated antibody
AV39532
Anti-NR1I2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV39738
Anti-NR1I2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1409956
Anti-NR1I2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411574
Anti-NR1I2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4504451
Anti-phospho-c-Jun (pSer243) antibody produced in rabbit, affinity isolated antibody
J2128 Anti-phospho-c-Jun (pSer63) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504368
Anti-phospho-c-Jun (pSer63) antibody produced in rabbit, affinity isolated antibody
J2253 Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504369
Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody
SAB4504452
Anti-phospho-c-Jun (pThr239) antibody produced in rabbit, affinity isolated antibody
SAB4504453
Anti-phospho-c-Jun (pThr91) antibody produced in rabbit, affinity isolated antibody
SAB4504454
Anti-phospho-c-Jun (pThr93) antibody produced in rabbit, affinity isolated antibody
SAB4504455
Anti-phospho-c-Jun (pTyr170) antibody produced in rabbit, affinity isolated antibody
SAB4300015
Anti-phospho-JUN (pSer243) antibody produced in rabbit, affinity isolated antibody
SAB4300002
Anti-phospho-JUN (pSer73) antibody produced in rabbit, affinity isolated antibody
SAB4300014
Anti-phospho-JUN (pThr239) antibody produced in rabbit, affinity isolated antibody
SAB4300011
Anti-phospho-JUN (pThr91) antibody produced in rabbit, affinity isolated antibody
SAB4300012
Anti-phospho-JUN (pThr93) antibody produced in rabbit, affinity isolated antibody
SAB4300013
Anti-phospho-JUN (pTyr170) antibody produced in rabbit, affinity isolated antibody
SAB4300200
Anti-phospho-MAPK14 (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4300201
Anti-phospho-MAPK14 (pTyr182) antibody produced in rabbit, affinity isolated antibody
P1491 Anti-phospho-p38 (pThr180/pTyr182) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504095
Anti-phospho-p38 MAPK (pThr179+Tyr181) antibody produced in rabbit, affinity isolated antibody
SAB4504497
Anti-phospho-p38 MAPK (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4504498
Anti-phospho-p38 MAPK (pTyr182) antibody produced in rabbit, affinity isolated antibody