EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Dual catalysis. Single-electron transmetalation in organoboron cross-coupling by photoredox/nickel dual catalysis.
PMID 24903560
Abstract
The routine application of C(sp3)-hybridized nucleophiles in cross-coupling reactions remains an unsolved challenge in organic chemistry. The sluggish transmetalation rates observed for the preferred organoboron reagents in such transformations are a consequence of the two-electron mechanism underlying the standard catalytic approach. We describe a mechanistically distinct single-electron transfer-based strategy for the activation of organoboron reagents toward transmetalation that exhibits complementary reactivity patterns. Application of an iridium photoredox catalyst in tandem with a nickel catalyst effects the cross-coupling of potassium alkoxyalkyl- and benzyltrifluoroborates with an array of aryl bromides under exceptionally mild conditions (visible light, ambient temperature, no strong base). The transformation has been extended to the asymmetric and stereoconvergent cross-coupling of a secondary benzyltrifluoroborate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
804215
[Ir{dFCF3ppy}2(bpy)]PF6
C34H18F16IrN4P
499625
Methyl 3-bromobenzoate, 98%
C8H7BrO2