EMAIL THIS PAGE TO A FRIEND
Journal of neurosurgery. Spine
Molecular characterization of chordoma xenografts generated from a novel primary chordoma cell source and two chordoma cell lines.
PMID 24905390
Abstract
Chordoma cells can generate solid-like tumors in xenograft models that express some molecular characteristics of the parent tumor, including positivity for brachyury and cytokeratins. However, there is a dearth of molecular markers that relate to chordoma tumor growth, as well as the cell lines needed to advance treatment. The objective in this study was to isolate a novel primary chordoma cell source and analyze the characteristics of tumor growth in a mouse xenograft model for comparison with the established U-CH1 and U-CH2b cell lines. Primary cells from a sacral chordoma, called "DVC-4," were cultured alongside U-CH1 and U-CH2b cells for more than 20 passages and characterized for expression of CD24 and brachyury. While brachyury is believed essential for driving tumor formation, CD24 is associated with healthy nucleus pulposus cells. Each cell type was subcutaneously implanted in NOD/SCID/IL2Rγ(null) mice. The percentage of solid tumors formed, time to maximum tumor size, and immunostaining scores for CD24 and brachyury (intensity scores of 0-3, heterogeneity scores of 0-1) were reported and evaluated to test differences across groups. The DVC-4 cells retained chordoma-like morphology in culture and exhibited CD24 and brachyury expression profiles in vitro that were similar to those for U-CH1 and U-CH2b. Both U-CH1 and DVC-4 cells grew tumors at rates that were faster than those for U-CH2b cells. Gross tumor developed at nearly every site (95%) injected with U-CH1 and at most sites (75%) injected with DVC-4. In contrast, U-CH2b cells produced grossly visible tumors in less than 50% of injected sites. Brachyury staining was similar among tumors derived from all 3 cell types and was intensely positive (scores of 2-3) in a majority of tissue sections. In contrast, differences in the pattern and intensity of staining for CD24 were noted among the 3 types of cell-derived tumors (p < 0.05, chi-square test), with evidence of intense and uniform staining in a majority of U-CH1 tumor sections (score of 3) and more than half of the DVC-4 tumor sections (scores of 2-3). In contrast, a majority of sections from U-CH2b cells stained modestly for CD24 (scores of 1-2) with a predominantly heterogeneous staining pattern. This is the first report on xenografts generated from U-CH2b cells in which a low tumorigenicity was discovered despite evidence of chordoma-like characteristics in vitro. For tumors derived from a primary chordoma cell and U-CH1 cell line, similarly intense staining for CD24 was observed, which may correspond to their similar potential to grow tumors. In contrast, U-CH2b tumors stained less intensely for CD24. These results emphasize that many markers, including CD24, may be useful in distinguishing among chordoma cell types and their tumorigenicity in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB3228
Anti-Cytokeratin 5 Antibody, 8, 100 µg, This Anti-Cytokeratin 5 Antibody, 8 is validated for use in FC, WB, IC, IH(P) for the detection of Cytokeratin 5.
SAB3701411 Anti-Human (κ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701417 Anti-Human (λ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW20082F Anti-Human IgA antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW20087F Anti-Human Kappa Light Chain antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA059479
Anti-KRT5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410739
Anti-KRT5 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
CKOZFN21103 CompoZr® Knockout ZFN Kit, Human TBX1 (NM_005992)
CKOZFND21103 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human TBX1 (NM_005992)
K0129
Kanamycin solution from Streptomyces kanamyceticus, 10 mg/mL in 0.9% NaCl, sterile-filtered, BioReagent, suitable for cell culture
SAB5300265 Monoclonal Anti-Cytokeratin 5 antibody produced in mouse, clone 1E1, ascites fluid
SAB5300267
Monoclonal Anti-Cytokeratin 5 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB4100508
Monoclonal Anti-KRT5 antibody produced in mouse, clone KRT5-A541, culture supernatant
SAB1402245
Monoclonal Anti-KRT5 antibody produced in mouse, clone 1A12, purified immunoglobulin, buffered aqueous solution