EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Tension force-induced ATP promotes osteogenesis through P2X7 receptor in osteoblasts.
PMID 24905552
Abstract
Orthodontic tooth movement induces alveolar bone resorption and formation by mechanical stimuli. Force exerted on the traction side promotes bone formation. Adenosine triphosphate (ATP) is one of the key mediators that respond to bone cells by mechanical stimuli. However, the effect of tension force (TF)-induced ATP on osteogenesis is inadequately understood. Accordingly, we investigated the effect of TF on ATP production and osteogenesis in MC3T3-E1 cells. Cells were incubated in the presence or absence of P2X7 receptor antagonist A438079, and then stimulated with or without cyclic TF (6% or 18%) for a maximum of 24 h using Flexercell Strain Unit 3000. TF significantly increased extracellular ATP release compared to control. Six percent TF had maximum effect on ATP release compared to 18% TF and control. Six percent TF induced the expression of Runx2 and Osterix. Six percent TF also increased the expression of extracellular matrix proteins (ECMPs), ALP activity, and the calcium content in ECM. A438079 blocked the stimulatory effect of 6% TF on the expression of Runx2, Osterix and ECMPs, ALP activity, and calcium content in ECM. This study indicated that TF-induced extracellular ATP is released in osteoblasts, suggesting that TF-induced ATP promotes osteogenesis by autocrine action through P2X7 receptor in osteoblasts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1000
1-Naphthol, ReagentPlus®, ≥99%
C10H8O
31097
1-Naphthol, PESTANAL®, analytical standard
C10H8O
N2780
1-Naphthol, BioXtra, ≥99%
C10H8O
33420
1-Naphthol, puriss. p.a., Reag. Ph. Eur., ≥99% (GC)
C10H8O
70440
1-Naphthol, puriss., for fluorescence, ≥99.0% (GC)
C10H8O
AV36678
Anti-RUNX2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36679
Anti-RUNX2 (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB1412189
ANTI-RUNX2 antibody produced in mouse, clone 6E1, purified immunoglobulin, buffered aqueous solution
SAB1412247
ANTI-RUNX2 antibody produced in mouse, clone 4E5, ascites fluid, buffered aqueous solution
SAB1412248
ANTI-RUNX2 antibody produced in mouse, clone 4D7, purified immunoglobulin, buffered aqueous solution
SAB1412249
ANTI-RUNX2 antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1412665
ANTI-RUNX2 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1412666
ANTI-RUNX2 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1412667
ANTI-RUNX2 antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
SAB1412668
ANTI-RUNX2 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
HPA022040
Anti-RUNX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106220
Anti-RUNX2 antibody produced in rabbit, affinity isolated antibody
WH0000860M1
Monoclonal Anti-RUNX2 antibody produced in mouse, clone 1D8, purified immunoglobulin, buffered aqueous solution
AMAB90591
Monoclonal Anti-RUNX2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0232, purified immunoglobulin, buffered aqueous glycerol solution
SAB1403638
Monoclonal Anti-RUNX2 antibody produced in mouse, clone 4D5, purified immunoglobulin, buffered aqueous solution
AMAB90594
Monoclonal Anti-RUNX2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0235, purified immunoglobulin, buffered aqueous glycerol solution