EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Cell cycle-dependent regulation of the RNA-binding protein Staufen1.
PMID 24906885
Abstract
Staufen1 (Stau1) is a ribonucleic acid (RNA)-binding protein involved in the post-transcriptional regulation of gene expression. Recent studies indicate that Stau1-bound messenger RNAs (mRNAs) mainly code for proteins involved in transcription and cell cycle control. Consistently, we report here that Stau1 abundance fluctuates through the cell cycle in HCT116 and U2OS cells: it is high from the S phase to the onset of mitosis and rapidly decreases as cells transit through mitosis. Stau1 down-regulation is mediated by the ubiquitin-proteasome system and the E3 ubiquitin ligase anaphase promoting complex/cyclosome (APC/C). Stau1 interacts with the APC/C co-activators Cdh1 and Cdc20 via its first 88 N-terminal amino acids. The importance of controlling Stau155 levels is underscored by the observation that its overexpression affects mitosis entry and impairs proliferation of transformed cells. Microarray analyses identified 275 Stau1(55)-bound mRNAs in prometaphase cells, an early mitotic step that just precedes Stau1 degradation. Interestingly, several of these mRNAs are more abundant in Stau155-containing complexes in cells arrested in prometaphase than in asynchronous cells. Our results point out for the first time to the possibility that Stau1 participates in a mechanism of post-transcriptional regulation of gene expression that is linked to cell cycle progression in cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB2502098
Anti-PCNA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502099
Anti-PCNA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701819
Anti-pcna antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV03018
Anti-PCNA antibody produced in rabbit, IgG fraction of antiserum
HPA030521
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA030522
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030523
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB4502103
Anti-PCNA antibody produced in rabbit, affinity isolated antibody
F2456
Fluorescein (free acid), Dye content 95 %
C20H12O5
EHU106031 MISSION® esiRNA, esiRNA human CDC20 (esiRNA1)
WH0000891M1
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1C8, purified immunoglobulin, buffered aqueous solution
SAB5300329 Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1B10, ascites fluid
SAB5300330
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 5G6, ascites fluid
WH0005111M2
Monoclonal Anti-PCNA antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
SAB1404178
Monoclonal Anti-PCNA antibody produced in mouse, clone S1, purified immunoglobulin, buffered aqueous solution
SAB4100556
Monoclonal Anti-PCNA antibody produced in mouse, clone PCNA -A509, culture supernatant
P9172
Pyronin Y, for NA electrophoresis
C17H19ClN2O
83200
Pyronin Y, for microscopy (Bot., Fl., Hist.)
C17H19ClN2O
213519
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
C17H19ClN2O
PLA0079
Rabbit anti-PCNA Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0080
Rabbit anti-PCNA Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.