EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
Effects of the compounds resveratrol, rutin, quercetin, and quercetin nanoemulsion on oxaliplatin-induced hepatotoxicity and neurotoxicity in mice.
PMID 24908156
Abstract
Oxaliplatin (OXA) is a platinum compound widely used in the treatment of some solid tumors, especially colorectal cancer. Despite its usefulness, oxaliplatin-associated neurotoxicity represents the main dose-limiting factor of this drug, and until now, there is no suitable treatment. Chemotherapy with oxaliplatin also increases the rate of developing hepatic damages with inflammatory activity, termed chemotherapy-associated steatohepatitis (CASH). In the present study, we aimed to compare the effects of a series of antioxidant compounds on simultaneous development of oxaliplatin-induced hepato- and neurotoxicity in mice. Mice BALB/c were treated with oxaliplatin for 6 weeks, 10 mg/kg, intraperitoneally, resulting in mechanical allodynia and hepatic steatosis. We administered the following antioxidant compounds--rutin (RT) (20 mg/kg), resveratrol (RVS) (100 mg/kg), quercetin (QT) (20 mg/kg), and quercetin nanoemulsion (NQT) (20 mg/kg)--daily by gavage to BALB/c, and N-acetylcysteine (NAC) was used as positive control. Treatments with RSV, RUT, or NQT were able to prevent mechanical allodynia when compared to the OXA group, and this effect was associated with decreased c-Fos immunopositivity in the lumbar spinal cord. Regarding the effects on steatohepatitis, RVS, QT, and NQT almost completely reversed the mean liver weight increase induced by OXA. In accordance with these previous data, histological evaluation indicated attenuation of all features of hepatic steatosis evaluated in RSV, RUT, QT, and NQT groups. These compounds were able to reduce the immunopositivity for the apoptosis marker caspase-3. On the other hand, only QT and NQT treatments were able to reduce neutrophil migration measured by myeloperoxidase (MPO) activity. These results suggest that the compounds tested, RSV, RUT, QT, and NQT, would be useful for the clinical treatment of neuro- and hepatoxicity induced by oxaliplatin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000271
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
C8H14N2O4Pt