EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Raman spectroscopy applied to the horizontal methods ISO 6579:2002 to identify Salmonella spp. in the food industry.
PMID 24908409
Abstract
Food safety is a major concern for suppliers in the food chain to ensure the safety of their products. The identification procedure requested by norms is tedious, and it often requires systematic controls and qualified staff to perform the necessary analyses. Raman spectroscopy offers new opportunities to rapidly and efficiently ascertain the presence of pathogens in samples. Nevertheless, this technique requires a standardized procedure to be applied in the industrial context. Our study shows that the variability between spectral fingerprints is related to the physiological state of the microbial species and the growth phase of the bacteria plays a crucial role in its identification by Raman spectroscopy. To improve the discrimination between closely related bacterial species, a procedure based on the selection of bacterial spectra in the exponential growth phase was proposed. Different ways to introduce Raman spectroscopy in the ISO 6579:2002 standards are also proposed from the entire process to a shorter protocol. In the latter case, the identification of bacterial colonies after the selective enrichment step was proposed with the advantages of this path in terms of simplicity and rapidity (analysis time is reduced up to 50 h from the 100 h required by the standard). The protocol validated using six food categories from industrial partners have presented a good correlation by confirmation with other laboratory classical methods. In the future, this procedure could be introduced to the control system of the food production chain with a reliable database for various microorganisms encountered in this field.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27066
Chromium, coating quality, 99.95%, grit
Cr
374849
Chromium, chips, 99.995% trace metals basis
Cr
255610
Chromium, chips, thickness ~2 mm, 99.5%
Cr
229563
Chromium, chunks, ≥99.99% trace metals basis
Cr
266264
Chromium, powder, 99.5%, −100 mesh
Cr
266299
Chromium, powder, ≥99% trace metals basis, −325 mesh
Cr
749052
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr