EMAIL THIS PAGE TO A FRIEND
Tuberculosis (Edinburgh, Scotland)
Increased frequency of ILT2-expressing CD56(dim)CD16(+) NK cells correlates with disease severity of pulmonary tuberculosis.
PMID 24909369
Abstract
The functional role of ILT2 in anti-tuberculosis immunity remains to be elucidated. In this study, we investigated expression and functions of ILT2 on NK cells during TB infection. The frequency of CD56(dim)CD16(+) NK cells that expressed ILT2 was significantly elevated in patients with active pulmonary TB as compared with tuberculin-positive healthy controls (pxa0