EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
High-throughput tandem mass spectrometry multiplex analysis for newborn urinary screening of creatine synthesis and transport disorders, Triple H syndrome and OTC deficiency.
PMID 24909877
Abstract
Creatine synthesis and transport disorders, Triple H syndrome and ornithine transcarbamylase deficiency are treatable inborn errors of metabolism. Early screening of patients was found to be beneficial. Mass spectrometry analysis of specific urinary biomarkers might lead to early detection and treatment in the neonatal period. We developed a high-throughput mass spectrometry methodology applicable to newborn screening using dried urine on filter paper for these aforementioned diseases. A high-throughput methodology was devised for the simultaneous analysis of creatine, guanidineacetic acid, orotic acid, uracil, creatinine and respective internal standards, using both positive and negative electrospray ionization modes, depending on the compound. The precision and accuracy varied by <15%. Stability during storage at different temperatures was confirmed for three weeks. The limits of detection and quantification for each biomarker varied from 0.3 to 6.3 μmol/l and from 1.0 to 20.9 μmol/l, respectively. Analyses of urine specimens from affected patients revealed abnormal results. Targeted biomarkers in urine were detected in the first weeks of life. This rapid, simple and robust liquid chromatography/tandem mass spectrometry methodology is an efficient tool applicable to urine screening for inherited disorders by biochemical laboratories.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G11608
Guanidineacetic acid, 99%
C3H7N3O2
O2750
Orotic acid, ≥98% (titration), anhydrous
C5H4N2O4