EMAIL THIS PAGE TO A FRIEND
Prenatal diagnosis
A new marker set that identifies fetal cells in maternal circulation with high specificity.
PMID 24912661
Abstract
Fetal cells from the maternal circulation (FCMBs) have the potential to replace cells from amniotic fluid or chorionic villi in a diagnosis of common chromosomal aneuploidies. Good markers for enrichment and identification are lacking. Blood samples from 78 normal pregnancies were used for testing the marker-set CD105 and CD141 for fetal cell enrichment. Fetal cell candidates were subsequently stained by a cocktail of cytokeratin antibodies, and the gender of the fetal cells was explored by fluorescence in situ hybridization (FISH) of the X and Y chromosomes. Fetal cell candidates could be detected in 91% of the samples, and in 85% of the samples, it was possible to obtain X and Y chromosomal FISH results for gender determination. The concordance between gender determined by FISH on fetal cells in maternal blood and gender found at birth reached 100% if three or more fetal cells with FISH signals could be found in a sample. The marker set identifies fetal cells with specificity high enough to make cell-based noninvasive prenatal diagnosis realistic.