EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Determination of sulfadiazine in phosphate- and DOC-rich agricultural drainage water using solid-phase extraction followed by liquid chromatography-tandem mass spectrometry.
PMID 24912991
Abstract
Trace levels of the veterinary antibiotic compound sulfadiazine (SDZ) can be determined in agricultural drainage water samples with this new method. Optimized sample pretreatment and solid-phase extraction was combined with liquid chromatography coupled to tandem mass spectrometry (SPE LC-MS/MS) using positive electrospray ionization. The linear dynamic range for the LC-MS/MS was assessed from 5 μg/L to 25 mg/L with a 15-point calibration curve displaying a coefficient of correlation r(2) = 0.9915. Agricultural drainage water spiked at a concentration of 25 ng/L gave recoveries between 63 and 98 % (relative standard deviation 15 %), while at 10 ng/L, it showed a lower recovery of 32 % (relative standard deviation 47 %). The final SPE LC-MS/MS method had a limit of detection (LOD)(Method) and a limit of quantification (LOQ)(Method) of 7.5 and 23 ng/L agricultural drainage water, respectively. Determination of SDZ, spiked at a realistic concentration of 50 μg/L, in artificial drainage water (ADW) containing common and high levels of phosphate (0.05, 0.5, and 5 mg/L) gave recoveries between 70 and 92 % (relative standard deviation 7.4-12.9 %). Analysis of the same realistic concentration of SDZ in ADW, spiked with common and high levels of dissolved organic carbon (2, 6, and 15 mg/L) confirmed the possible adaptation of a tandem solid-phase extraction (strong anion exchange (SAX)-hydrophilic-lipophilic balance (HLB)) followed by liquid chromatography-tandem mass spectrometry methodology. Recoveries obtained ranged from 104 to 109 % (relative standard deviation 2.8-5.2 %). The new methods enable determination of the veterinary antibiotic compound SDZ in agricultural drainage water from field experiments and monitoring schemes for phosphate- and dissolved organic carbon (DOC)-rich water samples in intensive farming areas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5080
Calcium chloride dihydrate, BioXtra, ≥99.0%
CaCl2 · 2H2O
C7902
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
CaCl2 · 2H2O
C2536
Calcium chloride dihydrate, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
CaCl2 · 2H2O
C3306
Calcium chloride dihydrate, for molecular biology, ≥99%
CaCl2 · 2H2O
21097
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
CaCl2 · 2H2O
21098
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
CaCl2 · 2H2O
21102
Calcium chloride dihydrate, tested according to Ph.Eur.
CaCl2 · 2H2O
27102
Chromoionophore XI, Selectophore
C38H48O5
C1909
Citric acid monohydrate, ACS reagent, ≥99.0%
C6H8O7 · H2O
C7129
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
C6H8O7 · H2O
C0706
Citric acid monohydrate, BioXtra, ≥99.5%
C6H8O7 · H2O
C2219000
Citric acid monohydrate, European Pharmacopoeia (EP) Reference Standard
C6H8O7 · H2O
58221
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
ClK
60126
Potassium chloride, tested according to Ph.Eur.
ClK
PHR1329
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClK
60137 Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl
ClK
60135
Potassium chloride solution, BioUltra, ~3 M in H2O
ClK
60134
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
ClK
60136
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
ClK
60138
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
ClK