EMAIL THIS PAGE TO A FRIEND
European cells & materials
Performance of different three-dimensional scaffolds for in vivo endochondral bone generation.
PMID 24913441
Abstract
In the context of skeletal tissue development and repair, endochondral ossification has inspired a new approach to regenerate bone tissue in vivo using a cartilage intermediate as an osteoinductive template. The aim of this study was to investigate the behavior of mesenchymal stem cells (MSCs) in regard to in vitro cartilage formation and in vivo bone regeneration when combined with different three-dimensional (3D) scaffold materials, i.e., hydroxyapatite/tricalcium phosphate (HA/TCP) composite block, polyurethane (PU) foam, poly(lactic-co-glycolic acid)/poly(ε-caprolactone) electrospun fibers (PLGA/PCL) and collagen I gel. To this end, rat MSCs were seeded on these scaffolds and chondrogenically differentiated in vitro for 4 weeks followed by in vivo subcutaneous implantation for 8 weeks. Nonetheless, the quality and maturity of in vivo ectopic bone formation appeared to be scaffold/material-dependent. Eight weeks of implantation was not sufficient to ossify the entire PLGA/PCL constructs, albeit a comprehensive remodeling of the cartilage had occurred. For HA/TCP, PU and collagen I scaffolds, more mature bone formation with rich vascularity and marrow stroma development could be observed. These data suggest that chondrogenic priming of MSCs in the presence of different scaffold materials allows the establishment of reliable templates for generating functional endochondral bone tissue in vivo without using osteoinductive growth factors. The morphology and maturity of bone formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74440
(−)-Nopol, purum, ≥98.0% (sum of enantiomers, GC)
C11H18O
N9389
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2765
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2640
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N4645
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
C6H4NNa2O6P · 6H2O
71768
4-Nitrophenyl phosphate disodium salt hexahydrate, for enzyme immunoassay, ≥99.0% (enzymatic)
C6H4NNa2O6P · 6H2O
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
P0380
L-Proline, ReagentPlus®, ≥99% (HPLC)
C5H9NO2
P5607
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
C5H9NO2
P8865
L-Proline, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO2
1568506
L-Proline, United States Pharmacopeia (USP) Reference Standard
C5H9NO2
81709
L-Proline, BioUltra, ≥99.5% (NT)
C5H9NO2
93693
L-Proline, certified reference material, TraceCERT®
C5H9NO2
W331902
L-Proline, 99%, FCC, FG
C5H9NO2
PHR1332
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
C5H9NO2
M9140
Methylene blue, certified by the Biological Stain Commission
C16H18ClN3S · xH2O
03978
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
C16H18ClN3S
1808
Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
C16H18ClN3S
319112
Methylene Blue solution, 0.05 wt. % in H2O
C16H18ClN3S
77515
Methylene Blue solution, for microscopy
C16H18ClN3S
50484
Methylene Blue solution, for microbiology
C16H18ClN3S
66725
Methylene Blue solution alkaline according to Loeffler, according to Loeffler
C16H18ClN3S
72438
N,N-Dimethylformamide, analytical standard
C3H7NO
72586
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
NNaO2
72587
Nitrite ion standard solution, 0.01 M NO2-, for ion-selective electrodes
NNaO2
P4744
Phosphatase substrate, powder
C6H4NNa2O6P · 6H2O
S0942
Phosphatase substrate, 5 mg tablets
C6H4NNa2O6P · 6H2O
P5994
Phosphatase substrate, 40 mg tablets
C6H4NNa2O6P · 6H2O
P5744
Phosphatase substrate, 40 mg capsules
C6H4NNa2O6P · 6H2O
P5869
Phosphatase substrate, 100 mg capsules
C6H4NNa2O6P · 6H2O