EMAIL THIS PAGE TO A FRIEND
Talanta
Comparison between solid phase microextraction (SPME) and hollow fiber liquid phase microextraction (HFLPME) for determination of extractables from post-consumer recycled PET into food simulants.
PMID 24913857
Abstract
Hollow fiber liquid phase microextraction (HFLPME) and solid phase microextraction (SPME) methods for pre-concentration of contaminants (toluene, benzophenone, tetracosane and chloroform) in food simulants were investigated. For HFLPME 1-heptanol, 2-octanone and dibutyl-ether were studied as extracting solvents. Analysis by gas chromatography coupled to mass spectrometry (GC-MS), flame ionization (GC-FID) and electron capture detectors (GC-ECD) were carried out. In addition, the methods were employed to evaluate the safety in use of a PET material after the recycling process (comprising washing, extrusion and solid state polymerization (SSP)) through extractability studies of the contaminants using 10% (v/v) ethanol in deionized water and 3% (w/v) acetic acid in deionized water as food simulants in different conditions: 10 days at 40°C and 2h at 70°C. The HFLPME preconcentration method provided increased sensitivity when compared to the SPME method and allowed to analyze concentration levels below 10 µg surrogate per kg food simulant. The results of the extractability studies showed considerable reductions after the extrusion and SSP processes and indicated the compliance with regulations for using recycled PET in contact with food.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72954
1-Heptanol, analytical standard
C7H16O
51778
1-Heptanol, ≥99.5% (GC)
C7H16O
51790
1-Heptanol, purum, ≥99.0% (GC)
C7H16O
H2805
1-Heptanol, 98%
C7H16O
02479
2-Octanone, analytical standard
C8H16O
O4709
2-Octanone, reagent grade, 98%
C8H16O
W280208
2-Octanone, ≥98%, FG
C8H16O
W280220
2-Octanone, natural, 98%, FG
C8H16O
427551
Benzophenone, purified by sublimation, ≥99%
C13H10O
239852
Benzophenone, ReagentPlus®, ≥99%
C13H10O
B9300
Benzophenone, ReagentPlus®, 99%
C13H10O
W213403
Benzophenone, ≥99%, FCC, FG
C13H10O
B3634
Benzophenone
C13H10O
12750
Benzophenone, purum, ≥99.0% (GC)
C13H10O
442842
Benzophenone, analytical standard
C13H10O
Y0000647
Benzophenone, European Pharmacopoeia (EP) Reference Standard
C13H10O
PHR1204
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
C13H10O
271454
Dibutyl ether, anhydrous, 99.3%
C8H18O
34462
Dibutyl ether, puriss. p.a., low in aromatic compounds, ≥99.5% (GC)
C8H18O
34470
Dibutyl ether, purum, ≥99.0% (GC)
C8H18O
110280
Dibutyl ether, ReagentPlus®, ≥99%
C8H18O
W254800
Heptyl alcohol, ≥97%, FCC, FG
C7H16O
W254820
Heptyl alcohol, natural, 98%
C7H16O
50296
Melting point standard 47-49°C, analytical standard
C13H10O
73664
Mettler-Toledo Calibration substance ME 18870, Benzophenone, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C13H10O
87089
Tetracosane, analytical standard
C24H50
T8752
Tetracosane, 99%
C24H50
442707
Tetracosane, analytical standard
C24H50