EMAIL THIS PAGE TO A FRIEND
Biomaterials
Cardiac RNAi therapy using RAGE siRNA/deoxycholic acid-modified polyethylenimine complexes for myocardial infarction.
PMID 24917027
Abstract
Inflammatory response in myocardial ischemia-reperfusion injury plays a critical role in ventricular remodeling. To avoid deleterious effects of overwhelming inflammation, we blocked the expression of receptor for advanced glycation end-products (RAGE), a key mediator of the local and systemic inflammatory responses, via RNAi mechanism. Herein, a facial amphipathic deoxycholic acid-modified low molecular weight polyethylenimine (DA-PEI) was used as a siRNA delivery carrier to myocardium. The DA-PEI conjugate formed a stable complex with siRNA via electrostatic and hydrophobic interactions. The siRAGE/DA-PEI formulation having negligible toxicity could enhance intracellular delivery efficiency and successfully suppress RAGE expression both inxa0vitro and inxa0vivo. Furthermore, the cardiac administration of siRAGE/DA-PEI reduced apoptosis and inflammatory cytokine release, subsequently led to attenuation of left ventricular remodeling in rat myocardial infarction model. The potential therapeutic effects of RAGE gene silencing on myocardial ischemia-reperfusion injury may suggest that the siRAGE/DA-PEI delivery system can be considered as a promising strategy for treating myocardial infarction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8877
2,3,5-Triphenyltetrazolium chloride, ≥95%
C19H15ClN4
36650
DCC, puriss., ≥99.0% (GC)
C13H22N2