EMAIL THIS PAGE TO A FRIEND
Science signaling
T cell receptor-dependent activation of mTOR signaling in T cells is mediated by Carma1 and MALT1, but not Bcl10.
PMID 24917592
Abstract
Signaling to the mechanistic target of rapamycin (mTOR) regulates diverse cellular processes, including protein translation, cellular proliferation, metabolism, and autophagy. Most models place Akt upstream of the mTOR complex, mTORC1; however, in T cells, Akt may not be necessary for mTORC1 activation. We found that the adaptor protein Carma1 [caspase recruitment domain (CARD)-containing membrane-associated protein 1] and at least one of its associated proteins, the paracaspase MALT1 (mucosa-associated lymphoid tissue lymphoma translocation protein 1), were required for optimal activation of mTOR in T cells in response to stimulation of the T cell receptor (TCR) and the co-receptor CD28. However, Bcl10, which binds to Carma1 and MALT1 to form a complex that mediates signals from the TCR to the transcription factor NF-κB (nuclear factor κB), was not required. The catalytic activity of MALT1 was required for the proliferation of stimulated CD4+ T cells, but not for early TCR-dependent activation events. Consistent with an effect on mTOR, MALT1 activity was required for the increased metabolic flux in activated CD4+ T cells. Together, our data suggest that Carma1 and MALT1 play previously unappreciated roles in the activation of mTOR signaling in T cells after engagement of the TCR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401063
Anti-CD69 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401064
Anti-CD69 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA002044
Anti-PSMC4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005471
Anti-PSMC4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105008
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2105009
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300484
Anti-RPS6 (Ab-235) antibody produced in rabbit, affinity isolated antibody
SAB1300250
Anti-RPS6 (N-term) antibody produced in rabbit, Ig fraction of antiserum, buffered aqueous solution
HPA031153
Anti-RPS6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107629
Anti-RPS6 antibody produced in rabbit, affinity isolated antibody
SAB1401065 Monoclonal Anti-CD69 antibody produced in mouse, clone 4H3, purified immunoglobulin, buffered aqueous solution
SAB4700219 Monoclonal Anti-CD69 antibody produced in mouse, clone FN50, purified immunoglobulin, buffered aqueous solution
WH0005704M1
Monoclonal Anti-PSMC4 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
WH0006194M1
Monoclonal Anti-RPS6 antibody produced in mouse, clone 3H1-F2, purified immunoglobulin, buffered aqueous solution