PloS one
Chondrogenesis of infrapatellar fat pad derived adipose stem cells in 3D printed chitosan scaffold.
PMID 24918443
Abstract
Infrapatellar fat pad adipose stem cells (IPFP-ASCs) have been shown to harbor chondrogenic potential. When combined with 3D polymeric structures, the stem cells provide a source of stem cells to engineer 3D tissues for cartilage repair. In this study, we have shown human IPFP-ASCs seeded onto 3D printed chitosan scaffolds can undergo chondrogenesis using TGFβ3 and BMP6. By week 4, a pearlescent, cartilage-like matrix had formed that penetrated the top layers of the chitosan scaffold forming a 'cap' on the scaffold. Chondrocytic morphology showed typical cells encased in extracellular matrix which stained positively with toluidine blue. Immunohistochemistry demonstrated positive staining for collagen type II and cartilage proteoglycans, as well as collagen type I. Real time PCR analysis showed up-regulation of collagen type II, aggrecan and SOX9 genes when IPFP-ASCs were stimulated by TGFβ3 and BMP6. Thus, IPFP-ASCs can successfully undergo chondrogenesis using TGFβ3 and BMP6 and the cartilage-like tissue that forms on the surface of 3D-printed chitosan scaffold may prove useful as an osteochondral graft.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid
C6H8O6
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
16911
Hydrogen peroxide solution, for ultratrace analysis
H2O2
95321
Hydrogen peroxide solution, ≥30%, for trace analysis
H2O2
88597
Hydrogen peroxide solution, 3%, for microbiology
H2O2
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
95212
L-Ascorbic acid, tested according to Ph.Eur.
C6H8O6
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se
204307
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Se
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
HT-SE101
Selenium, pellets, < 5mm, ≥99.999%
Se
GF66395212
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Se