EMAIL THIS PAGE TO A FRIEND
Neuroreport
Involvement of extracellular factors in maintaining self-renewal of neural stem cell by nestin.
PMID 24918459
Abstract
Nestin knockout leads to embryonic lethality and self-renewal deficiency in neural stem cells (NSCs). However, how nestin maintains self-renewal remains uncertain. Here, we used the dosage effect of nestin in heterozygous mice (Nes+/-) to study self-renewal of NSCs. With existing extracellular signaling in vivo or in vitro, nestin levels do not affect proliferation ability or apoptosis when compared between Nes+/- and Nes+/+ NSCs. However, self-renewal ability of Nes+/- NSCs is impaired when plated at a low cell density and completely lost at a clonal density. This deficiency in self-renewal at a clonal density is rescued using a medium conditioned by Nes+/+ NSCs. In addition, the Akt signaling pathway is altered at low density and reversed by conditioned medium. Our data show that secreted factors contribute toward maintaining self-renewal of NSCs by nestin, potentially through Akt signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9564
DAPI, dilactate, ≥98% (HPLC)
C16H15N5 · 2C3H6O3