PloS one
Characterization of the xiamenmycin biosynthesis gene cluster in Streptomyces xiamenensis 318.
PMID 24919072
Abstract
Xiamenmycin (1) is a prenylated benzopyran derivative with anti-fibrotic activity. To investigate the genetic basis of xiamenmycin biosynthesis, we performed genome mining in the xiamenmycin-producing Streptomyces xiamenensis wild-type strain 318 to identify a candidate gene cluster. The complete gene cluster, consisting of five genes, was confirmed by a series of gene inactivations and heterologous expression. Based on bioinformatics analyses of each gene and feeding experiments, we found that the structure of an intermediate xiamenmycin B (3) accumulated in a ximA inactivation mutant, allowing us to propose a biosynthetic pathway. All five of the genes in the pathway were genetically and biochemically characterized. XimA was biochemically characterized as an ATP-dependent amide synthetase, catalyzing an amide bond formation in the presence of ATP as the final step in Xiamenmycin biosynthesis. The Km value of XimA was determined to be 474.38 µM for the substrate xiamenmycin B. These studies provide opportunities to use genetic and chemo-enzymatic methods to create new benzopyran derivatives as potential therapeutic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54630
4-Hydroxybenzoic acid, puriss., ≥99.0% (T)
C7H6O3
240141
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
C7H6O3
PHR1048
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
H20059
4-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
W398608
4-Hydroxybenzoic acid, ≥99%, FG
C7H6O3
N5755
β-Nicotinamide adenine dinucleotide phosphate hydrate
C21H28N7O17P3 · xH2O
N8035
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
C21H27N7NaO17P3
N8160
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
C21H27N7NaO17P3
C1200000
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
C11H12Cl2N2O5
31667
Chloramphenicol, VETRANAL, analytical standard
C11H12Cl2N2O5
23276
Chloramphenicol, tested according to Ph.Eur.
C11H12Cl2N2O5
T8250
D-Threonine, ≥98% (TLC)
C4H9NO3
D4287
γ,γ-Dimethylallyl pyrophosphate triammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥90% (TLC)
C5H12O7P2 · 3NH3
G6772
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
C10H20O7P2 · 3H3N
T8625
L-Threonine, reagent grade, ≥98% (HPLC)
C4H9NO3
T8441
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
C4H9NO3
T4071
L-Threonine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C4H9NO3
89179
L-Threonine, BioXtra, ≥99.5% (NT)
C4H9NO3
61506
L-Threonine, certified reference material, TraceCERT®
C4H9NO3
PHR1242
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H9NO3
T1340000
L-Threonine, European Pharmacopoeia (EP) Reference Standard
C4H9NO3
N8878
Nalidixic acid, ≥98%
C12H12N2O3
N5035
Nalidixic acid, meets USP testing specifications
C12H12N2O3
1451000
Nalidixic acid, United States Pharmacopeia (USP) Reference Standard
C12H12N2O3
97023
Nalidixic acid, analytical standard
C12H12N2O3
N0050000
Nalidixic acid, European Pharmacopoeia (EP) Reference Standard
C12H12N2O3