PloS one
Epigenetic regulations of immediate early genes expression involved in memory formation by the amyloid precursor protein of Alzheimer disease.
PMID 24919190
Abstract
We previously demonstrated that APP epigenetically regulates Egr1 expression both in cultured neurons and in vivo. Since Egr1 is an immediate early gene involved in memory formation, we wondered whether other early genes involved in memory were regulated by APP and we studied molecular mechanisms involved. By comparing prefrontal (PF) cortex from wild type (APP+/+) and APP knockout mice (APP-/-), we observed that APP down regulates expression of four immediate early genes, Egr1, c-Fos, Bdnf and Arc. Down regulation of Egr1, c-Fos and Bdnf transcription resulted from a decreased enrichment of acetylated histone H4 on the corresponding gene promoter. Further characterization of H4 acetylation at Egr1 and c-Fos promoters revealed increased acetylation of H4K5 and H4K12 residues in APP-/- mice. Whereas APP affected Egr1 promoter activity by reducing access of the CREB transcription factor, its effect on c-Fos appeared to depend on increased recruitment of HDAC2 histone deacetylase to the gene promoter. The physiological relevance of the epigenetic regulation of Egr1 and c-Fos gene transcription by APP was further analyzed following exposure of mice to novelty. Although transcription of Egr1 and c-Fos was increased following exposure of APP+/+ mice to novelty, such an induction was not possible in APP-/- mice with a high basal level of expression of these immediate early genes. Altogether, these results demonstrate that APP-mediated regulation of c-Fos and Egr1 by different epigenetic mechanisms is needed for their induction during exposure to novelty.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500441
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500442
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500443
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500444
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500445
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4300519
Anti-CREB1 (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300333
Anti-CREB1 (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB4300546
Anti-CREB1 (Ab-142) antibody produced in rabbit, affinity isolated antibody
AV37464
Anti-CREB1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV01026
Anti-CREB1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV31264
Anti-CREB1 antibody produced in rabbit, IgG fraction of antiserum
HPA019150
Anti-CREB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103540
Anti-CREB1 antibody produced in rabbit, affinity isolated antibody
SAB4300412
Anti-HDAC2 (Ab-394) antibody produced in rabbit, affinity isolated antibody
HPA011727
Anti-HDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32618
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB2104633
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
H3159
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H2663
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture
AV38791
Anti-HTATIP antibody produced in rabbit, affinity isolated antibody
SAB2103488
Anti-HTATIP antibody produced in rabbit, affinity isolated antibody
SAB2104133
Anti-HTATIP, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1407003
Anti-KAT5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA016953
Anti-KAT5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106496
Anti-KAT5 antibody produced in rabbit, affinity isolated antibody
SAB4500117
Anti-TIP60 antibody produced in rabbit, affinity isolated antibody
SAB4500118
Anti-TIP60 antibody produced in rabbit, affinity isolated antibody
SAB4500119
Anti-TIP60 antibody produced in rabbit, affinity isolated antibody
C8977 Monoclonal Anti-CREB antibody produced in rabbit, clone 48H2, purified immunoglobulin, buffered aqueous glycerol solution
WH0001385M1
Monoclonal Anti-CREB1 antibody produced in mouse, clone 3F1-1B2, purified immunoglobulin, buffered aqueous solution